Search
With bench space at a premium, we offer a variety of microtube rack solutions to organize your laboratory work space while conducting, transporting, and storing your experiments. Pick the rack that fits your needs best.
| Nalgene Microtube Rack | Nalgene Floating Microtube Rack | Thermo Scientific Flipstrip Rack | Thermo Scientific Reversible Rack | Thermo Scientific Flipper Rack | Thermo Scientific 4-way Flipper |
| Autoclavable. Rugged racks that hold up in harsh lab environments. | Ideal for genetic, microbiology, and immunology labs. Freezer to water bath service for thawing and incubating microtubes. Square racks for turbulent waters, round racks fit in 400mL and 1000mL Nalgene beakers. | Supports transfer from 1.5mL/2.0mL tubes into 0.2mL tubes; positions for 12 larger tubes horizontally across the top of the rack and 12 strips of 0.2mL tubes vertically below each of the larger tubes. Flip the rack over and the other side holds 96 0.5mL tubes. | The little flipper. Double-sided - holds 0.2mL PCR tubes on one side and 0.5mL PCR tubes on the other. | The big flipper. Double-sided—holds 0.5mL tubes on one side and 1.5mL/2.0mL on the other. | Flip it your way. Four orientations convert this rack between four tube sizes ranging including 0.5mL microtubes; 1.5/2.0mL microtubes; 15mL conicals; 50mL conicals |
![]() | ![]() | ![]() | ![]() | ![]() | ![]() |
| Resmer Manufacturing Technology | Polypropylene | Recycled plastic | Recycled plastic | Polypropylene | Polypropylene |
| No lid | No lid | With clear lid | With clear lid | With clear lid | No lid |
| Handle | Handle | Side rail handles | Side rail handles | Side rail handles | No handle |
| Autoclavable | Do not autoclave; temp range –70°C to 100°C | Do not autoclave | Do not autoclave | Do not autoclave | Do not autoclave |
| Single sided | Single sided | Double sided | Double sided | Double sided | 4 sided |
| Tube locations keyed with numbers and letters | Tube locations keyed with numbers and letters | Tube locations keyed with numbers and letters | Tube locations keyed with numbers and letters | Tube locations keyed with numbers and letters | Tube locations keyed with numbers and letters |
| Stackable | Nest for storage | Stackable | Stackable | Stackable | Stackable |
| Does not float in water | Floats | Does not float in water | Does not float in water | Does not float in water | Does not float in water |
| Holds 32, 1.5mL microtubes | Holds 1.0, 1.2, 1.5 and 2.0mL tubes | Holds 12 1.5 or 2.0mL tubes and 96 0.2mL tubes. Other side holds 96 0.5mL tubes | Holds 96 0.2mL on one side or 96 0.5mL tubes on the other side | Holds 96 0.5mL on one side or 96 1.5/2.0mL tubes on the other side | Holds 32 0.5, 1.5, 2.0mL tubes or 12 15mL tubes or 4 50mL tubes |
| 4 x 6 array | 4 x 4 array 20 tubes (round rack) 8 tubes (round rack) | 8 x 12 array plus 12 on one side | 8 x 12 array | 8 x 12 array | Various arrays |
| White | White/clear | Fluorescent green, pink, orange & assorted | Fluorescent green, pink, orange & assorted | Fluorescent green, pink, orange | Fluorescent green, pink, orange |
Receive a Certificate of Compliance and Certificate(s) of Irradiation (if applicable) on purchased Thermo Scientific Nalgene and Nunc products.
Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.