Anti-Horse Secondary Antibodies
Our anti-horse secondary antibodies are affinity-purified polyclonal antibodies with well-characterized specificity for horse immunoglobulin class. They are available in host species including goat and rabbit, and in the following formats:
- Unlabeled antibodies
- Enzyme conjugates (alkaline phosphatase (AP) or horseradish peroxidase (HRP))
- Fluorescent conjugates (Fluorescein (FITC)
- Biotin
Our secondary antibodies are available as whole IgG. Each of these antibodies is affinity purified using its target antigen coupled to an agarose support. This affinity purification eliminates nonspecific antibodies, resulting in high specificity and low background signal. Conjugated antibodies are affinity purified before labeling.
How to find a secondary antibody
Step 1: Use the tool at the top of the page to filter by target, application, target species
Step 2: View more filters to narrow results by host species, conjugate and other criteria
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.
Resources
Product selection guides
Learning and support
For Research Use Only. Not for use in diagnostic procedures.