On this page:
A single-use alternative for traditional stirred-tank mixing
The Thermo Scientific HyPerforma DS 300 Single-Use Mixer (S.U.M.) is built upon the proven legacy design of the S.U.M, but optimized for lower-cost, high throughput applications. The smaller footprint and lighter weight of the HyPerforma DS 300 S.U.M. offer mobility and flexibility in mixing applications not currently available in the market. The system utilizes a top-mounted mixing motor with an angular drive shaft. This design aims to eliminate vortices, support efficient mixing, and help provide more power.
HyPerforma DS 300 Single-Use Mixer
A cost-effective docking station solution, consisting of a mixing station and a plastic support container containing a BioProcess Container (BPC) or tank liners. Optional dollies are available to maneuver the support containers for all drums sizes. 
Support container and BPC size availability:
  • Top-drain support containers: 50, 100, 200, and 300 L
  • Bottom-drain support containers: 50, 100, and 200 L
  • Top-drain tank liners: 50–300 L
  • Closed-top 3D BPCs with top and bottom access ports: 50, 100, and 200 L
Features and applications
  • The docking station consists of a stainless-steel base with locking casters, an adjustable handle, a tethered hand-held control device, an electronic vertical lift mechanism with integrated height indicator, motor mount and motor, three sizes of drive shafts, and two adjustment tools—a spanner and a torque wrench. An adjustable-angle motor head and drum positioned are available as add-on options.
  • Supporting hardware optimized for mixing, storage, transport and discharge in a closed system to help minimize the risk of cross-contamination
  • Standard BPCs available for liquid/liquid and powder/liquid mixing
Reliable BPCs and consumables for every step in your process
The open-top, non-irradiated BPC tank liner with bottom drain is composed of CX3-9 single web, multi-layer plastic film, compatible with the HyPerforma DS 300 S.U.M. BPCs and liners come in open- and closed-top design and are manufactured using our proprietary CX5-14 and CX3-9 film that easily fit in many plastic drum sizes, so you can implement liners in the overall process of shipping and storing your liquid-liquid and powder-liquid bioprocess mixing applications.
Need to contact a specialist or get support for your single-use mixer?
Single-Use Bioprocessing Equipment and Automation
We are committed to delivering exceptional performance, reliability, and quality.
Single-Use Bioprocessing
Our innovative single-use technologies enable you to accelerate biological manufacturing processes to create flexibility and operate more dynamically within upstream and downstream workflows.
BioProcessing Containers and Nalgene Bottles
We offer high-quality storage, transport, and sampling solutions, including Thermo Scientific BioProcess Containers (BPCs) and Thermo Scientific Nalgene containers.
Speak to a specialist
Connect with a Bioprocessing Single-Use Technology representative.
Additional bioprocessing resources
Discover areas where you can cut costs and gain efficiency using our integrated calculator.
Explore our collection of information and insights to support your drug development journey.
Learn, evaluate, connect.
Connect with a Bioprocessing representative or request more info.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.