Search
With the same growth kinetics as laboratory-scale cell culture products, Nunc Cell Factory systems are a proven solution for large-scale production of cells, vaccines, and therapeutic proteins. Our hands-on expertise will help you accelerate productivity and ease the burden of regulatory compliance. This quality production cell culture platform reflects the rigorous standards that have made us the world's leading provider of cell culture products.
An innovative replacement for traditional cable ties with a 360-degree seal, the BioTitan Retention Device is designed to enhance the reliability and integrity of assemblies and lower particulate risk, with zero leaks reported since its 2021 launch. Now available on Nunc Cell Factory systems.
Nunc Cell Factory systems are a proven solution for research using adherent cell culture. These systems have the same growth kinetics as other laboratory-scale cell culture products.
Available in 1-layer to 40-layer formats, this solution enables consistent performance and enhanced productivity for scale-up.
The closed system Nunc Standard Cell Factory systems are built with standard components to help ensure quality as well as assurance of supply. This ready-to-use standard closed system helps you quickly develop and scale-up your adherent cell culture process.
Available in 2-layer and 10-layer formats, learn how you can get consistent, high-quality results with this ready-to-use closed system.
With true configured Nunc Cell Factory systems and equipment, advance your fully commercial-scale process with pre-assembled Cell Factory systems and automation equipment. Our commercial team will work with you to design a custom closed system and automation process that fit your specific needs.
Nunc Cell Factory systems are a proven solution for research using adherent cell culture. These systems have the same growth kinetics as other laboratory-scale cell culture products.
Available in 1-layer to 40-layer formats, this solution enables consistent performance and enhanced productivity for scale-up.
The closed system Nunc Standard Cell Factory systems are built with standard components to help ensure quality as well as assurance of supply. This ready-to-use standard closed system helps you quickly develop and scale-up your adherent cell culture process.
Available in 2-layer and 10-layer formats, learn how you can get consistent, high-quality results with this ready-to-use closed system.
With true configured Nunc Cell Factory systems and equipment, advance your fully commercial-scale process with pre-assembled Cell Factory systems and automation equipment. Our commercial team will work with you to design a custom closed system and automation process that fit your specific needs.
Enhance workflow productivity
To address the needs of your workflow, the portfolio offers the EasyFill-2 Cell Factory system. The EasyFill-2 system contains dual large port openings to accommodate faster rates of emptying and filling and increased system aeration. It has one large opening and a port opening for easy and secure tubing assemblies. The system features our certified Thermo Scientific Nunclon Delta cell culture surface, which is available on most Nunc cell culture products to ensure consistent performance from lot to lot and from format to format. The system also comes in multiple-train versions and are compatible with Cell Factory equipment to support process scale-up.
Maximize productivity with automation solutions. Develop a fully commercial-scale process using Nunc Cell Factory equipment options.
Let us help you customize your Nunc Cell Factory system to fit your specific needs. Use our guided checklist in this flyer to indicate desired layers, port size and locations, connector, vent filter, and tubing details.
Get started now
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.