Search
Determine the clarity of your sample due to particulate content and meet regulatory requirements when using a Thermo Scientific Orion AQUAfast Turbidity Meter. AQUAfast Turbidity meters feature white and infrared sources and are designed with convenience and portability in mind.
Take your turbidity meter to the source for on-site measuring. AQUAfast Turbidity Meters feature white and infrared sources allowing you to meet regulatory methods.
Advanced performance for any environment
| |
Reliable value anywhere you need it
|
Take your portable, IP 67 waterproof turbidity meters to the source for on-site measuring. Designed to help meet regulatory requirements with EPA 180.1 and ISO 7027 compliant models.
| Product | Description | |
|---|---|---|
| Orion AQUAfast AQ3010 Turbidity Meter | Learn more | |
Reliable value anywhere you need it • Infrared (IR) LED meets ISO 7027 and other global method requirements | ||
| Orion AQUAfast AQ4500 Turbidimeter | Learn more | |
Advanced performance for any environment • White LED light meets U.S. EPA 180.1 and other global method requirements | ||
Cat. No. | Description | For use with |
|
AC45ST | EPA compliant primary turbidity standard calibration, set of 5 | Orion AQUAfast AQ4500 | |
AC45FZ | 4000 NTU primary formazin turbidity standard, 475mL | Orion AQUAfast AQ4500, Orion AQUAfast AQ3010 | |
AC301S | ISO compliant polymer-based turbidity standard kit, set of 4 | Orion AQUAfast AQ3010 |
We offer the turbidity accessories you'll need to support your turbidimety meter, including vials, primary turbidity standards, Formazin turbidity standards, cleaning brushes, and more.
Cat. No. | Description | For use with |
|
AC2T24 | Replacement 24mm turbidity vials, 12/pack | AQ4500 | |
AC45S1 | Silicon oil and cloth | AQ4500 | |
AQ4CBL | RS232 cable for data download and software update | AQ4500 | |
AC3CBR | Vial cleaning brush, 5/pack | AQ4500 | |
AC3V25 | Replacement 25mm vials, 3/pack | AQ3010 | |
AC3SIL | Silicon oil and cloth | AQ3010 |
Take your portable, IP 67 waterproof turbidity meters to the source for on-site measuring. Designed to help meet regulatory requirements with EPA 180.1 and ISO 7027 compliant models.
| Product | Description | |
|---|---|---|
| Orion AQUAfast AQ3010 Turbidity Meter | Learn more | |
Reliable value anywhere you need it • Infrared (IR) LED meets ISO 7027 and other global method requirements | ||
| Orion AQUAfast AQ4500 Turbidimeter | Learn more | |
Advanced performance for any environment • White LED light meets U.S. EPA 180.1 and other global method requirements | ||
Cat. No. | Description | For use with |
|
AC45ST | EPA compliant primary turbidity standard calibration, set of 5 | Orion AQUAfast AQ4500 | |
AC45FZ | 4000 NTU primary formazin turbidity standard, 475mL | Orion AQUAfast AQ4500, Orion AQUAfast AQ3010 | |
AC301S | ISO compliant polymer-based turbidity standard kit, set of 4 | Orion AQUAfast AQ3010 |
We offer the turbidity accessories you'll need to support your turbidimety meter, including vials, primary turbidity standards, Formazin turbidity standards, cleaning brushes, and more.
Cat. No. | Description | For use with |
|
AC2T24 | Replacement 24mm turbidity vials, 12/pack | AQ4500 | |
AC45S1 | Silicon oil and cloth | AQ4500 | |
AQ4CBL | RS232 cable for data download and software update | AQ4500 | |
AC3CBR | Vial cleaning brush, 5/pack | AQ4500 | |
AC3V25 | Replacement 25mm vials, 3/pack | AQ3010 | |
AC3SIL | Silicon oil and cloth | AQ3010 |
Get answers to your top water analysis and water purification questions and see products in action. Learn about:
· pH & Multiparameter Benchtop & Portable Meters
· Electrodes (pH Electrodes, ISE, Conductivity, DO)
· Potentiometric Titrators
· Spectrophotometers
· Water Purification Systems