Search
The Attune™ NxT Acoustic Focusing Cytometer is a benchtop analyzer (Figure 1) that uses acoustic focusing—a revolutionary technology that precisely aligns cells using ultrasonic waves prior to interrogation with one or more lasers. This technology enables multicolor flow cytometric analysis with significantly greater collection rates and an improved ability to detect rare events without excess sample manipulation. The Attune™ NxT system offers:
![]() | Figure 1. Attune™ NxT Acoustic Focusing Cytometer with optional Attune™ NxT Autosampler, for processing up to 384 samples. |
With up to 4 lasers and 14 emission channels for multiparameter analysis, the Attune™ NxT Acoustic Focusing Cytometer was designed as a modular system to fit most experimental needs and lab budgets (Table 1). The Attune™ NxT cytometer can be configured to accommodate most common fluorophores used in flow cytometry, as well as to match the panels you are currently running. For example, with the optional violet and yellow lasers (in addition to the standard blue laser), the Attune™ NxT cytometer can be used to simultaneously detect multiple fluorescent proteins. Whether you configure your system now or upgrade later, the Attune™ NxT Acoustic Focusing Cytometer can grow with you and your research needs.
| Feature | Attune™ NxT Acoustic Focusing Cytometer |
|---|---|
| Size, weight | 40 x 58 x 43 cm (16 x 23 x 17 in), 29 kg (64 lb) |
| Electrical | 100–240 VAC, 50/60 Hz, <150 W |
| Excitation | Up to 4 lasers: Violet (405 nm, 50 mW), blue (488 nm, 50 mW), yellow (561 nm, 50 mW), red (637 nm, 100 mW) Laser profile: Flat-top laser requiring minimal alignment Flow cell: Quartz cuvette gel-coupled to 1.2 NA collection lens |
| Emission | Forward scatter: Photodiode detector with 488/10 nm bandpass filter Side scatter: PMT with 488/10 nm bandpass filter Emission filters: User-changeable, keyed filters |
| Fluidics | Sample rate: 12.5–1,000 μL/min Sample analysis volume: 20 μL–4 mL Sample tube size: 17 x 100 mm to 8.5 x 45 mm Sample delivery: Positive-displacement syringe pump Fluid storage: All fluids stored within instrument with active fluid level sensing (optional external tank available) |
| Performance | Data acquisition rate: Up to 35,000 events/sec Maximum event file: 20 million Particle size range: 0.5–50 μm Automated maintenance: ≤15 min startup and shutdown |
For Research Use Only. Not for use in diagnostic procedures.