Search
Introduction
Instructions are provided below for electrophoresis of the Novex® Pre-Cast Gels using the XCell SureLock™ Mini-Cell. For more information on the XCell SureLock™ Mini-Cell, refer to the manual (IM-9003). You may download this manual from our website or contact Technical Service.
If you are using any other mini-cell for electrophoresis, refer to the manufacturer’s recommendations.
Protocol using XCell SureLock™ Mini-Cell
Wear gloves and safety glasses when handling gels.
XCell SureLock™ Mini-Cell requires 200 ml for the Upper Buffer Chamber and 600 ml for the Lower Buffer Chamber.
See below for Electrophoresis Conditions.
Electrophoresis Conditions
Run your gels according to the following protocol:
Gel Type | Voltage | Expected Current* | Run Time |
Tris-Glycine Gels (SDS-PAGE)
|
125 V constant
|
Start: 30-40mA
End: 8-12 mA
|
90 minutes (dependent on gel type)
Run the gel until the bromophenol blue tracking dye reaches the bottom of the gel.
|
Tris-Glycine Gels (Native-PAGE)
|
125 V constant
|
Start: 6-12 mA
End: 3-6 mA
|
1-12 hours
|
*Expected start and end current values are stated for single gels.
Removing the Gel after Electrophoresis
The Novex® Tris-Glycine SDS or Native Sample Buffer (2X) and NuPAGE® Reducing Agent (10X) are available from Thermo Fisher Scientific.
| Reagent | Denatured Sample | Native Sample |
|---|---|---|
| Sample | x µl | x µl |
| Tris-Glycine SDS Sample Buffer (2X) | 5 µl | -- |
| Tris-Glycine Native Sample Buffer (2X) | -- | |
| Deionized Water | to 5 µl | to 5 µl |
| Total Volume | 10 µl | 10 µl |
Running Reduced and Non-Reduced Samples
For optimal results, we do not recommend running reduced and non-reduced samples on the same gel.
If you do choose to run reduced and non-reduced samples on the same gel, do not run reduced and non-reduced samples in adjacent lanes. The reducing agent may have a carry-over effect on the non-reduced samples if they are in close proximity.
Heating Samples
Heating the sample at 100° C in SDS containing buffer results in proteolysis (Kubo, 1995). We recommend heating samples for denaturing electrophoresis (reduced or non-reduced) at 85°C for 2-5 minutes for optimal results.
Do not heat the samples for non-denaturing (native) electrophoresis.
Recommended Buffers
The recommended running buffer and sample buffer for each Novex® Pre-Cast Gel is listed in the table below. You will prepare your sample in the appropriate sample buffer so the final concentration of the sample buffer will be 1X. You will need to dilute the running buffer to 1X final concentration before use.
See recipes if you are making your own buffers.
| Gel Type | Running Buffer | Sample Buffer |
|---|---|---|
| Tris-Glycine Gel (SDS-PAGE) | Tris-Glycine SDS Running Buffer (10X) | Tris-Glycine SDS Sample Buffer (2X) |
Reducing Agent
The NuPAGE® Reducing Agent contains 500 mM dithiothreitol (DTT) at a 10X concentration in a ready-to-use, stabilized liquid form and is used to prepare samples for reducing gel electrophoresis.
ß-mercaptoethanol can be used with the Novex® gels at a final concentration of 2.5%. Choice of the reducing agent is a matter of preference and either DTT or ß-mercaptoethanol can be used. We recommend adding the reducing agent to the sample within an hour of loading the gel.
Avoid storing reduced samples for long periods even if they are frozen. This will result in the reoxidation of samples during storage and produce inconsistent results.
Running Reduced and Non-Reduced Samples
For optimal results, we do not recommend running reduced and non-reduced samples on the same gel.
If you do choose to run reduced and non-reduced samples on the same gel, do not run reduced and non-reduced samples in adjacent lanes. The reducing agent may have a carry-over effect on the non-reduced samples if they are in close proximity.
Novex® Tris-Glycine SDS or Native Running Buffer (10X) is available from Thermo Fisher Scientific.
1. Prepare 1000 ml of 1X Tris-Glycine SDS or Native Running Buffer using Novex® Tris-Glycine SDS or Native Running Buffer (10X) as follows:
| Novex® Tris-Glycine SDS or Native Running Buffer (10X) | 100 ml |
| Deionized Water | 900 ml |
| Total Volume | 1000 ml |
2. Mix thoroughly. Use this buffer to fill the Upper and Lower Buffer Chambers of the XCell SureLock™ Mini-Cell for electrophoresis.
See recipes for the Novex® Tris-Glycine SDS and Native Running Buffers, if you are preparing the running buffers
Introduction
The molecular weight of a protein can be determined based upon its relative mobility by constructing a standard curve with protein standards of known molecular weights.
The protein mobility in SDS-PAGE gels is dependent on the:
An identical molecular weight standard may have slightly different mobility resulting in different apparent molecular weight when run in different SDS-PAGE buffer systems. If you are using the Novex® protein molecular weight standards, see the apparent molecular weights of these standards on the Novex® Pre-Cast Gels listed to determine an apparent molecular weight of your protein.
Protein Secondary Structure
When using SDS-PAGE for molecular weight determination, slight deviations from the calculated molecular weight of a protein (calculated from the known amino acid sequence) can occur due to the retention of varying degrees of secondary structure in the protein, even in the presence of SDS. This phenomenon is observed in highly organized secondary structures (such as collagens, histones, or highly hydrophobic membrane proteins) and in peptides, where the effect of local secondary structure and amino acid side chains becomes magnified relative to the total size of the peptide.
Buffer Systems
Slight differences in protein mobilities also occur when the same proteins are run in different SDS-PAGE buffer systems. Each SDS-PAGE buffer system has a different pH, which affects the charge of a protein and its binding capacity for SDS. The degree of change in protein mobility is usually small in natural proteins but more pronounced with “atypical” or chemically modified proteins such as pre-stained standards.
Assigned Apparent Molecular Weights
The apparent molecular weight values currently provided with the Novex® molecular weight standards were derived from the construction of a calibration curve in the Tris-Glycine SDS-PAGE System. We have now calculated and assigned apparent molecular weights for the Novex® protein standards in several buffer systems. Remember to use the one that matches your gel for the most accurate calibration of your protein.
The following charts summarize the approximate molecular weight values for the Novex® protein molecular weight standards when run in different buffer systems. You may generate calibration curves in your lab with any other manufacturer’s standards.
MultiMark® Multi-Colored Standard | Tris-Glycine Gels (4-20%) |
Myosin
|
250 kDa
|
Phosphorylase B
|
148 kDa
|
Glutamic Dehydrogenase
|
60 kDa
|
Carbonic Anhydrase
|
42 kDa
|
Myoglobin (Blue)
|
30 kDa
|
Myoglobin (Red)
|
22 kDa
|
Lysozyme
|
17 kDa
|
Aprotinin
|
6 kDa
|
Insulin
|
4 kDa
|
Mark 12™ Unstained Standard | Tris-Glycine Gels (4-20%) |
Myosin
|
200 kDa
|
ß-Galactosidase
|
116.3 kDa
|
Phosphorylase B
|
97.4 kDa
|
Bovine Serum Albumin
|
66.3 kDa
|
Glutamic Dehydrogenase
|
55.4 kDa
|
Lactate Dehydrogenase
|
36.5 kDa
|
Carbonic Anhydrase
|
31 kDa
|
Trypsin Inhibitor
|
21.5 kDa
|
Lysozyme
|
14.4 kDa
|
Aprotinin
|
6 kDa
|
Insulin B Chain
|
Unresolved Insulin
|
SeeBlue® Pre-Stained Standard | Tris-Glycine Gel (4-20%) |
Myosin
|
250 kDa
|
BSA
|
98 kDa
|
Glutamic Dehydrogenase
|
64 kDa
|
Alcohol Dehydrogenase
|
50 kDa
|
Carbonic Anhydrase
|
36 kDa
|
Myoglobin
|
30 kDa
|
Lysozyme
|
16 kDa
|
Aprotinin
|
6 kDa
|
Insulin
|
4 kDa
|
SeeBlue® Plus2 Pre-Stained Standard | Tris-Glycine Gel (4-20%) |
Myosin
|
250 kDa
|
Phosphorylase B
|
148 kDa
|
BSA
|
98 kDa
|
Glutamic Dehydrogenase
|
64 kDa
|
Alcohol Dehydrogenase
|
50 kDa
|
Carbonic Anhydrase
|
36 kDa
|
Myoglobin
|
22 kDa
|
Lysozyme
|
16 kDa
|
Aprotinin
|
6 kDa
|
Insulin
|
4 kDa
|
Problem | Cause | Solution |
Run taking longer time
|
Running buffer too dilute
|
Make fresh running buffer as described in this manual and avoid adjusting the pH of the 1X running buffer.
|
Low or no current during the run
|
Incomplete circuit
|
|
Faint shadow or “ghost” band below the expected protein band
|
Ghost bands are caused due to a slight lifting of the gel from the cassette resulting in trickling of some sample beyond its normal migration point. Gel lifting off the cassette is caused due to:
|
|
Streaking of proteins
|
|
|
|
| |
|
| |
|
| |
Bands in the outer lane of the gel are curving upwards
|
|
|
|
| |
|
| |
Bands in the outside lanes of the gel “smiling”
|
Expired gels used causing the acrylamide to break down in the gel
|
Avoid using gels after the expiration date. Use fresh gels.
|
Bands are running as U shape rather than a flat band
|
Samples are loaded on the gel and not electrophoresed immediately resulting in sample diffusion
|
Load samples on to the gel immediately before electrophoresis.
|
Bands appear to be “funneling” or getting narrower as they progress down the gel
|
Proteins are over-reduced causing the proteins to be negatively charged and repel each other.
|
Reduce the proteins using DTT or ß-mercaptoethanol as described.
|
Dumbbell shaped bands after electrophoresis
|
Loading a large volume of sample causing incomplete stacking of the entire sample. This effect is intensified for larger proteins
|
Load the appropriate volume of sample per well as described. If your sample is too dilute, concentrate the sample using salt precipitation or ultrafiltration.
|
Tris-Glycine SDS Running Buffer
The Tris-Glycine SDS Running Buffer is available from Thermo Fisher Scientific.
25 mM Tris Base
192 mM Glycine
0.1% SDS
pH 8.3
1. To prepare 1000 ml of 10 X Tris-Glycine SDS Running Buffer, dissolve the following reagents to 900 ml ultrapure water:
Tris Base 29 g
Glycine 144 g
SDS 10 g
2. Mix well and adjust the volume to 1000 ml with ultrapure water.
3. Store at room temperature. The buffer is stable for 6 months when stored at room temperature.
4. For electrophoresis, dilute this buffer to 1X with water. The pH of the 1X solution is 8.3. Do not use acid or base to adjust the pH.
Tris-Glycine Native Running Buffer
The Tris-Glycine Native Running Buffer is available from Thermo Fisher Scientific.
25 mM Tris base
192 mM Glycine
pH 8.3
1. To prepare 1000 ml of 10 X Tris-Glycine Native Running Buffer, dissolve the following reagents to 900 ml ultrapure water:
Tris Base 29 g
Glycine 144 g
2. Mix well and adjust the volume to 1000 ml with ultrapure water.
3. Store at room temperature. The buffer is stable for 6 months when stored at room temperature.
4. For native electrophoresis, dilute this buffer to 1X with water. The pH of the 1X solution is 8.3. Do not use acid or base to adjust the pH.
Tris-Glycine SDS Sample Buffer
The Tris-Glycine SDS Sample Buffer is available from Thermo Fisher Scientific.
63 mM Tris HCl
10% Glycerol
2% SDS
0.0025% Bromophenol Blue
pH 6.8
1. To prepare 10 ml of 2X Tris-Glycine SDS Sample Buffer, mix the following reagents :
0.5 M Tris-HCl, pH 6.8 2.5 ml
Glycerol 2 ml
10% (w/v) SDS 4 ml
0.1% (w/v) Bromophenol Blue 0.5 ml
2. Adjust the volume to 10 ml with ultrapure water.
3. Store at +4° C. The buffer is stable for 6 months when stored at +4° C.
Tris-Glycine Native Sample Buffer
The Tris-Glycine Native Sample Buffer is available from Thermo Fisher Scientific.
100 mM Tris HCl
10% Glycerol
0.0025% Bromophenol Blue
pH 8.6
1. To prepare 10 ml of 2X Tris-Glycine Native Sample Buffer, mix the following reagents :
0.5 M Tris HCl, pH 8.6 4 ml
Glycerol 2 ml
0.1% (w/v) Bromophenol Blue 0.5 ml
2. Adjust the volume to 10 ml with ultrapure water.
3. Store at +4° C. The buffer is stable for 6 months when stored at +4° C.
Tris-Glycine Transfer Buffer
The Tris-Glycine Transfer Buffer is available from Thermo Fisher Scientific.
12 mM Tris Base
96 mM Glycine
pH 8.3
1. To prepare 500 ml of 25 X Tris-Glycine Transfer Buffer, dissolve the following reagents in 400 ml ultrapure water:
Tris Base 18.2 g
Glycine 90 g
2. Mix well and adjust the volume to 500 ml with ultrapure water.
3. Store at room temperature. The buffer is stable for 6 months when stored at room temperature.
4. For blotting, dilute this buffer as described. The pH of the 1X solution is 8.3. Do not use acid or base to adjust the pH.
Compatibility
The size of a Novex® Pre-Cast Gel is 10 x 10 cm (gel size is 8 x 8 cm). We recommend using the XCell SureLock™ Mini-Cell for the electrophoresis of Novex® Pre-Cast Gels to obtain optimal and consistent performance. Novex® Pre-Cast Gels are compatible with most other mini-cells designed for electrophoresis of 10 cm (h) x 10 cm (w) gel cassettes.
Staining Novex® Pre-Cast Gels
The Novex® Pre-Cast Gels are compatible with most silver staining protocols. We recommend using the SilverQuest™ Silver Staining Kit or the SilverXpress® Silver Staining Kit for silver staining of Novex® Gels.
The Novex® Pre-Cast Gels are compatible with any of the standard Coomassie® staining procedures. The protocols that are accelerated by heat are preferable as heat serves as a “fix” for proteins, especially smaller peptides. The SimplyBlue™ SafeStain and Novex® Colloidal Coomassie® Blue Staining Kit are recommended for staining Novex® Gels.
Specifications
Gel Matrix: Acrylamide/Bisacrylamide
Gel Thickness: 1.0 mm or 1.5 mm
Gel Size: 8 cm x 8 cm
Cassette Size: 10 cm x 10 cm
Cassette Material: Styrene Copolymer (recycle code 7)
Sample Well Configuration 1, 5, 9, 10, 12, 15-well, 2D -well, and IPG well
Recommended Loading Volumes
The recommended loading volumes and protein load per band by the detection method are provided in the table below.
Note: The 9-well gels are compatible with any eight-channel pipettors used for loading samples from 96-well plates. An additional lane is included for loading protein molecular weight standard. 