DPBS (10X), no calcium, no magnesium
Gibco™

DPBS (10X), no calcium, no magnesium

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
14200075500 mL
1420016610 x 500 mL
Catalog number 14200075
Price (KRW)
77,000
Online offer
Ends: 31-Dec-2026
85,000
Save 8,000
Each
Quantity:
500 mL
Customize this product
Price (KRW)
77,000
Online offer
Ends: 31-Dec-2026
85,000
Save 8,000
Each

Dulbecco's phosphate-buffered saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. Formulations without calcium and magnesium are required for rinsing chelators from the culture before cell dissociation. Concentrated forms of Gibco™ DPBS require pH adjustment and filtration at the time of preparation (see protocol for details).

We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications.

This DPBS is modified as follows:

Without: Calcium, Magnesium, Phenol Red

The complete formulation is available.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Chemical Name or MaterialDPBS
ColorClear
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality2630 to 3000 mOsm/kg
Recommended StorageStorage conditions: 15°C to 30°C, Shipping conditions: Room temperature, Shelf life: 36 months from date of manufacture
SterilitySterile
Sterilization MethodSterile-filtered
Concentration10X
For Use With (Application)Mammalian Cell Culture
GradeCell Culture
Physical FormLiquid
Product LineGibco
Quantity500 mL
Solution TypeCleaning, Storage, pH Buffer
pH6.7 to 7.0
Unit SizeEach

Frequently asked questions (FAQs)

Do you offer an alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538)?

We do not have a direct alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538). However, we do have alternative primary and secondary antibodies, as well as reagents, that can be used for trilineage differentiation. Please note that we have not internally validated the use of all these reagents together.

We recommend the following primary antibodies:

Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4 (asm-1)) (Cat. No. MA5-11547)

alpha-Fetoprotein Monoclonal Antibody (AFP3) (Cat. No. 14-6583-80)

beta-3 Tubulin Monoclonal Antibody (2G10) (Cat. No. MA1-118)

We recommend the following secondary antibodies:

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 555 (Cat. No. A21137)

Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 594 (Cat. No. A21135)

https://www.thermofisher.com/antibody/product/Goat-anti-Mouse-IgG1-Cross-Adsorbed-Secondary-Antibody-Polyclonal/A-21121

We recommend using the following reagents:

https://www.thermofisher.com/order/catalog/product/88-8824-00

NucBlue Fixed Cell ReadyProbes Reagent (DAPI) (Cat. No. R37606)

Blocker BSA (Cat. No. 37520)

DPBS (10X), no calcium, no magnesium (Cat. No. 14200075)

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Citations & References (1)

Citations & References
Abstract
Isolation of multipotent neural stem or progenitor cells from both the dentate gyrus and subventricular zone of a single adult mouse.
Authors:Guo W, Patzlaff NE, Jobe EM, Zhao X,
Journal:Nat Protoc
PubMed ID:23080272
In adult mammals, the subventricular zone (SVZ) of the lateral ventricles and the subgranular zone of the dentate gyrus (DG) show ongoing neurogenesis, and multipotent neural stem or progenitor cells (NSCs) in these two regions exhibit different intrinsic properties. However, investigation of the mechanisms underlying such differences has been limited ... More