DPBS (10X), no calcium, no magnesium, 500 mL - FAQs
DPBS (10X), no calcium, no magnesium, 500 mL - FAQs
View additional product information for DPBS (10X), no calcium, no magnesium - FAQs (14200075, 14200166)
3 product FAQs found
Do you offer an alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538)?
We do not have a direct alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538). However, we do have alternative primary and secondary antibodies, as well as reagents, that can be used for trilineage differentiation. Please note that we have not internally validated the use of all these reagents together.
We recommend the following primary antibodies:
Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4 (asm-1)) (Cat. No. MA5-11547)
alpha-Fetoprotein Monoclonal Antibody (AFP3) (Cat. No. 14-6583-80)
beta-3 Tubulin Monoclonal Antibody (2G10) (Cat. No. MA1-118)
We recommend the following secondary antibodies:
Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 555 (Cat. No. A21137)
Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 594 (Cat. No. A21135)
https://www.thermofisher.com/antibody/product/Goat-anti-Mouse-IgG1-Cross-Adsorbed-Secondary-Antibody-Polyclonal/A-21121
We recommend using the following reagents:
https://www.thermofisher.com/order/catalog/product/88-8824-00
NucBlue Fixed Cell ReadyProbes Reagent (DAPI) (Cat. No. R37606)
Blocker BSA (Cat. No. 37520)
DPBS (10X), no calcium, no magnesium (Cat. No. 14200075)
Can I get a specific value for the conductivity for your DPBS products?
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Are your DPBS products DNase/RNase-free? Can I use them for RNA work?
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.