Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs
Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs
View additional product information for Goat anti-Mouse IgG2a Cross-Adsorbed Secondary Antibody, Alexa Fluor™ 555 - FAQs (A21137)
2 product FAQs found
Do you offer an alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538)?
We do not have a direct alternative for the discontinued 3-Germ Layer ICC Kit (Cat. No. A25538). However, we do have alternative primary and secondary antibodies, as well as reagents, that can be used for trilineage differentiation. Please note that we have not internally validated the use of all these reagents together.
We recommend the following primary antibodies:
Alpha-Smooth Muscle Actin Monoclonal Antibody (1A4 (asm-1)) (Cat. No. MA5-11547)
alpha-Fetoprotein Monoclonal Antibody (AFP3) (Cat. No. 14-6583-80)
beta-3 Tubulin Monoclonal Antibody (2G10) (Cat. No. MA1-118)
We recommend the following secondary antibodies:
Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 555 (Cat. No. A21137)
Goat anti-Mouse IgG2a Cross-Adsorbed, Alexa Fluor 594 (Cat. No. A21135)
https://www.thermofisher.com/antibody/product/Goat-anti-Mouse-IgG1-Cross-Adsorbed-Secondary-Antibody-Polyclonal/A-21121
We recommend using the following reagents:
https://www.thermofisher.com/order/catalog/product/88-8824-00
NucBlue Fixed Cell ReadyProbes Reagent (DAPI) (Cat. No. R37606)
Blocker BSA (Cat. No. 37520)
DPBS (10X), no calcium, no magnesium (Cat. No. 14200075)
I have two primary antibodies from the same host species that I wish to use to label the same cells. How do I choose secondary antibodies?
If the two primary antibodies are from different isotypes (such as mouse IgG1 and IgG2a), there are isotype-specific secondary antibodies to choose from. The other option is to make direct conjugates of the primary antibodies through the use of antibody label kits, reactive dyes, or custom conjugations.