Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
PCR Plate, 96-well, low profile, non-skirted, black lettering
Thermo Scientific™

PCR Plate, 96-well, low profile, non-skirted, black lettering

The low profile Thermo Scientific PCR plate (Black Lettering) has shorter wells than the standard profile plate (AB-0600), decreasing theRead more
Have Questions?
Catalog NumberQuantity
AB0700L25 plates
Catalog number AB0700L
Price (USD)
151.65
Online exclusive
176.00
Save 24.35
Each
Quantity:
25 plates
Request bulk or custom format
Price (USD)
151.65
Online exclusive
176.00
Save 24.35
Each
The low profile Thermo Scientific PCR plate (Black Lettering) has shorter wells than the standard profile plate (AB-0600), decreasing the 'dead space' between the heated lid of the thermal cycler and the sample. This eliminates condensation on the side wall of the tube, preventing reduction in PCR volume and increasing the efficiency of the reaction. These low profile products are especially recommended for use with reaction volumes below 20 μL.

Features of low profile Thermo Scientific PCR plates

• Low profile to reduce dead space and to increase PCR efficiency
• Black Lettering facilitates sample tracking
• Available with black alpha-numeric lettering
• Improved access for liquid handling
• Maximum fill volume of 0.2 mL
• Available in assorted colors
• Compatible with standard multichannel pipettes.
• Cut corner: H12

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
PCR Plate, 96-well, low profile, non-skirted
PCR Plate, 96-well, low profile, non-skirted, blue
PCR Plate, 96-well, low profile, non-skirted, green
PCR Plate, 96-well, low profile, non-skirted, black
PCR Plate, 96-well, low profile, non-skirted, red
PCR Plate, 96-well, low profile, non-skirted, white
PCR Plate, 96-well, low profile, non-skirted, yellow
PCR Plate, 96-well, low profile, non-skirted, black lettering
PCR Plate, 96-well, non-skirted
PCR Plate, 96-well, non-skirted, white
PCR Plate, 96-well, non-skirted, blue
PCR Plate, 96-well, non-skirted, green
PCR Plate, 96-well, non-skirted, black
PCR Plate, 96-well, non-skirted, purple
PCR Plate, 96-well, non-skirted, red
PCR Plate, 96-well, non-skirted, yellow
PCR Plate, 96-well, non-skirted, black lettering
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 plates
Reaction SpeedLow profile
Skirt StyleNon-skirt
Volume (Metric) Well0.2 mL
Volume (Metric) Working0.1 mL
BarcodeNo Barcode
ColorClear, Clear
MaterialPolypropylene
No. of Wells96
Unit SizeEach