Thermo Scientific™
Selectris and Selectris X Imaging Filters
Catalog number: SELECTRIS
Thermo Scientific™
Selectris and Selectris X Imaging Filters
Catalog number: SELECTRIS
The Thermo Scientific™ Selectris and Selectris X Imaging Filters are post-column imaging filters that improve the contrast of TEM images, resulting in high-resolution structures up to atomic resolution. The zero-loss filtering of the Selectris Filters removes noise caused by inelastically scattered electrons, producing an increased signal-to-noise ratio (SNR) and better contrast. Designed for high stability, the zero-loss peak position of the Selectris Filters is insensitive to temperature variations in the environment, eliminating the need for frequent tuning.
Paired with the latest generation Thermo Scientific Falcon 4i Direct Electron Detector, Selectris Filters enable you to obtain high-resolution structures quickly, revealing new biological insights. To further increase the productivity and ease-of-use of your cryo-electron microscopy (cryo-EM) workflows for single particle analysis and cryo-electron tomography, these filters are fully integrated into Thermo Scientific EPU Software, Thermo Scientific Tomography Software, and our instrument operation software.
The Selectris and Selectris X Imaging Filters are available on Thermo Scientific Krios and Glacios Cryo-TEMs.
Learn more about Selectris and Selectris X Imaging Filters >
Catalog Number
SELECTRIS
Unit Size
Each
Price (USD)
Full specifications
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizePrice (USD)
SELECTRISFull specifications
EachRequest A Quote
Unit SizeEach
Showing 1 of 1
Key Benefits
Designed for stability and atomic resolution imaging
  • Narrow (<10 eV) zero-loss energy filtering offers enhanced contrast for higher resolution reconstructions
  • Sophisticated aberration correction, low image distortions, and uniform energy resolution over entire field of view
  • Minimized sensitivity to temperature variations
Straightforward operation
  • Fully integrated with Thermo Scientific instrument operation software, as well as EPU Software and Tomography Software for data collection
  • Automated filter tuning completed in minutes
Falcon 4i Direct Electron Detector
  • High detective quantum efficiency (DQE)
  • High speed, routinely capable of producing 300+ movies/hour
  • Built-in electron event representation (EER) for full temporal resolution (320 fps to disk) - no need for fractionating at time of acquisition
  • Super resolution without file size penalty