Talos F200X G2 Transmission Electron Microscope
The Thermo Scientific Talos F200X STEM is a scanning transmission electron microscope that combines outstanding high-resolution STEM and TEM imaging with industry-leading energy dispersive X-ray spectroscopy (EDS) signal detection. 2D/3D chemical characterization with compositional mapping is performed by 4 in-column SDD Super-X detectors with unique cleanliness. The Talos F200X scanning transmission electron microscope allows for the fastest and most precise EDS analysis in all dimensions, along with high-resolution TEM and STEM (HRTEM and HRSTEM) imaging with fast navigation for dynamic microscopy. The Talos F200X scanning transmission electron microscope also features reduced environmental sensitivity; the instrument enclosure moderates the impact of air pressure waves, air flows, and fine temperature variations in the TEM room.
STEM imaging and chemical analysis
The Talos F200X (S)TEM delivers fast, precise, quantitative characterization of nanomaterials in multiple dimensions. With innovative features designed to increase throughput, precision, and ease of use, the Talos F200X (S)TEM is ideal for advanced research and analysis in academic, government, semiconductor and industrial environments.
The need for large area correlative imaging at high resolution has recently increased as it allows researchers to preserve the context of their observations while also providing statistically robust data. Thermo Scientific Maps Software (enabled by Thermo Scientific Velox Software) automatically acquires an array of images across a sample and stitches them together to create one large final image. Image acquisition can even be performed unattended. The APW (Automated Particle Workflow) pack has all the benefits described in this section and adds unique processing on a dedicated processing PC with Thermo Scientific Avizo2D Software. You can get nanoparticle parameters like size, area, perimeter, shape, factor, contacts, etc., in an automated way. The fully automated and unattended software pack enables you to use the Talos F200X (S)TEM 24/7, get much better statistics and significantly improve the repeatability because operator bias is not present.
Align Genie Automation Software eases the learning curve for novice operators, reduces tensions in a multi-user environment, and improves time-to-data for the experienced operator.
Talos F200X G2 Transmission Electron Microscope features
Available with a wide range of high-resolution field emissions guns (FEG)
Choose high-brightness X-FEG, or ultra-high-brightness Cold Field Emission Gun (X-CFEG). X-CFEG combines the best (S)TEM imaging with the best energy resolution. 
Intuitive software
Thermo Scientific Velox Software offers fast and easy acquisition and analysis of multimodal data.
Faster time to chemical composition
Rapid, precise quantitative EDS analysis reveals nanoscale details in 2D and 3D with high cleanliness.
Better image data
High throughput STEM imaging with simultaneous, multiple signal detection delivers better contrast for high quality images.
Space for more
Add application-specific in situ sample holders for dynamic experiments.
High-quality (S)TEM images and accurate EDS
Acquire high-quality TEM or STEM images with the innovative and intuitive Velox Software user interface. Unique EDS absorption correction in Velox Software enables highly accurate quantification.  
Highly repeatable data collection
All daily TEM tunings, such as focus, eucentric height, beam shift, condenser aperture, beam tilt pivot points and rotation center are automated, ensuring you always start from optimum imaging conditions. Experiments can be repeated reproducibly, allowing you to focus on your research rather than on instrument operation.
Increased productivity
Ultra-stable column and remote operation with the SmartCam Camera and constant-power objective lenses for swift mode and high-voltage (HT) switches. Fast and easy switching for multi-user environments.
Specifications
Hoja de estilo para las especificaciones de la tabla de productos
HRTEM line resolution
  • ≤0.10 nm
STEM resolution
  • ≤0.16 nm (X-FEG)
  • ≤0.14 nm with 100 pA (X-CFEG)
Super-X EDS system
  • 4 SDD symmetric design, windowless, shutter-protected
Electron energy loss spectroscopy (EELS) energy resolution
  • ≤0.8 eV (X-FEG)
  • ≤0.3 eV (X-CFEG
Gun brightness at 200 kV
  • 1.8×109 A /cm2 srad (X-FEG)
  • 2.4×109 A /cm2 srad (X-CFEG)
Style Sheet for Komodo Tabs
Resources
Applications
Process Control_Thumb_274x180_144DPI
Control de proceso
La industria moderna exige un alto rendimiento con una calidad superior, un equilibrio que se mantiene a través de un control de procesos sólido. Las herramientas SEM y TEM con software de automatización exclusivo proporcionan información rápida y multiescala para la supervisión y la mejora de procesos.
Quality Control_Thumb_274x180_144DPI
Control de calidad
El control y garantía de calidad son esenciales en la industria moderna. Ofrecemos una gama de herramientas de EM y espectroscopía para el análisis multiescala y multimodal de defectos, lo que le permite tomar decisiones fiables e informadas para el control y la mejora de procesos.
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Investigación sobre materiales fundamentales
Se investigan nuevos materiales a escalas cada vez más pequeñas para lograr el máximo control de sus propiedades físicas y químicas. La microscopía electrónica proporciona a los investigadores información clave sobre una amplia variedad de características materiales a escala nanométrica.
pathfinding_thumb_274x180_144dpi
Desarrollo y trazabilidad de semiconductores
Microscopía electrónica avanzada, haz de iones enfocado y técnicas analíticas asociadas para identificar soluciones viables y métodos de diseño para la fabricación de dispositivos semiconductores de alto rendimiento.
yield_ramp_metrology_2_thumb_274x180
Metrología y rampa de producción
Ofrecemos capacidades analíticas avanzadas para el análisis de defectos, metrología y control de procesos, diseñadas para ayudar a aumentar la productividad y mejorar el rendimiento en una amplia gama de aplicaciones y dispositivos semiconductores.
Análisis de fallos de semiconductores
Análisis de fallos de semiconductores
Las estructuras de dispositivos semiconductores cada vez más complejas dan lugar a que existan más ubicaciones en las que se oculten los defectos inducidos por fallos. Nuestros flujos de trabajo de última generación le ayudarán a localizar y caracterizar los sutiles problemas eléctricos que afectan a la producción, al rendimiento y a la fiabilidad.
physical_characterization_thumb_274x180_144dpi
Caracterización física y química
La demanda continua de los consumidores impulsa la creación de dispositivos electrónicos más pequeños, más rápidos y más baratos. Su producción se basa en instrumentos y flujos de trabajo de alta productividad que generan imágenes, analizan y caracterizan una amplia gama de semiconductores y dispositivos de visualización.
Techniques
Espectroscopia de energía dispersiva
La espectroscopía de energía dispersiva (EDS) recopila información elemental detallada junto con imágenes de microscopía electrónica, proporcionando un contexto de composición esencial para las observaciones de EM. Con EDS, se puede determinar la composición química desde barridos de superficie rápidos y holísticos hasta átomos individuales.
Más información ›
Tomografía EDS en 3D
La investigación de materiales modernos depende cada vez más del análisis a nanoescala en tres dimensiones. La caracterización en 3D, incluidos los datos de composición para el contexto químico y estructural completo, es posible con EM en 3D y espectroscopia de rayos X dispersiva.
Más información ›
Análisis elemental EDS
EDS proporciona información de composición vital sobre las observaciones de microscopio electrónico. En concreto, nuestros exclusivos sistemas de detectores Super-X y Dual-X añaden opciones para mejorar el rendimiento y/o la sensibilidad, permitiendo optimizar la adquisición de datos para cumplir con sus prioridades de investigación.
Más información ›
Asignación elemental a escala atómica con EDS
El EDS de resolución atómica proporciona un contexto químico incomparable para el análisis de materiales al diferenciar la identidad elemental de los átomos individuales. Cuando se combina con TEM de alta resolución, es posible observar la organización precisa de los átomos en una muestra.
Más información ›
Espectroscopía de pérdida de energía de electrones
La investigación en ciencias de los materiales se beneficia de la EELS de alta resolución para una amplia gama de aplicaciones analíticas. Esto incluye asignación elemental de alto rendimiento, alta relación señal-ruido, así como sondeo de estados de oxidación y fonones de superficie.
Más información ›
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Más información ›
Análisis de partículas
El análisis de partículas juega un papel vital en la investigación de nanomateriales y el control de calidad. La resolución a escala nanométrica y la adquisición de imágenes superiores de microscopía electrónica se pueden combinar con software especializado para la rápida caracterización de polvos y partículas.
Más información ›
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.
Más información ›
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Adquisición de imágenes y análisis TEM de semiconductores
Los microscopios de electrones de transmisión de Thermo Fisher Scientific ofrecen imágenes y análisis de alta resolución de dispositivos semiconductores, lo que permite a los fabricantes calibrar conjuntos de herramientas, diagnosticar mecanismos de fallos y optimizar la producción rendimiento general del proceso.
Más información ›
Espectroscopia de energía dispersiva
La espectroscopía de energía dispersiva (EDS) recopila información elemental detallada junto con imágenes de microscopía electrónica, proporcionando un contexto de composición esencial para las observaciones de EM. Con EDS, se puede determinar la composición química desde barridos de superficie rápidos y holísticos hasta átomos individuales.
Más información ›
Tomografía EDS en 3D
La investigación de materiales modernos depende cada vez más del análisis a nanoescala en tres dimensiones. La caracterización en 3D, incluidos los datos de composición para el contexto químico y estructural completo, es posible con EM en 3D y espectroscopia de rayos X dispersiva.
Más información ›
Análisis elemental EDS
EDS proporciona información de composición vital sobre las observaciones de microscopio electrónico. En concreto, nuestros exclusivos sistemas de detectores Super-X y Dual-X añaden opciones para mejorar el rendimiento y/o la sensibilidad, permitiendo optimizar la adquisición de datos para cumplir con sus prioridades de investigación.
Más información ›
Asignación elemental a escala atómica con EDS
El EDS de resolución atómica proporciona un contexto químico incomparable para el análisis de materiales al diferenciar la identidad elemental de los átomos individuales. Cuando se combina con TEM de alta resolución, es posible observar la organización precisa de los átomos en una muestra.
Más información ›
Espectroscopía de pérdida de energía de electrones
La investigación en ciencias de los materiales se beneficia de la EELS de alta resolución para una amplia gama de aplicaciones analíticas. Esto incluye asignación elemental de alto rendimiento, alta relación señal-ruido, así como sondeo de estados de oxidación y fonones de superficie.
Más información ›
Experimentación in situ
La observación directa y en tiempo real de los cambios microestructurales con microscopía electrónica es necesaria para comprender los principios subyacentes de los procesos dinámicos como la recristalización, el crecimiento del grano y la transformación de fases durante el calentamiento, refrigeración y humectación.
Más información ›
Análisis de partículas
El análisis de partículas juega un papel vital en la investigación de nanomateriales y el control de calidad. La resolución a escala nanométrica y la adquisición de imágenes superiores de microscopía electrónica se pueden combinar con software especializado para la rápida caracterización de polvos y partículas.
Más información ›
Análisis de escala múltiple
Los novedosos materiales se deben analizar a una resolución cada vez mayor, manteniendo el contexto más amplio de la muestra. El análisis de escala múltiple permite la correlación de varias herramientas y modalidades de obtención de imágenes, tales como microTC de rayos X, DualBeam, PFIB láser, SEM y TEM.
Más información ›
Metrología de TEM
Las rutinas de metrología TEM avanzadas y automatizadas ofrecen una precisión significativamente mayor que los métodos manuales. Esto permite a los usuarios generar grandes cantidades de datos estadísticamente relevantes, con una especificidad de nivel subangstrom, sin fallos del operador.
Más información ›
Adquisición de imágenes y análisis TEM de semiconductores
Los microscopios de electrones de transmisión de Thermo Fisher Scientific ofrecen imágenes y análisis de alta resolución de dispositivos semiconductores, lo que permite a los fabricantes calibrar conjuntos de herramientas, diagnosticar mecanismos de fallos y optimizar la producción rendimiento general del proceso.
Más información ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Servicios de microscopía electrónica
Para garantizar un rendimiento óptimo del sistema, le proporcionamos acceso a una red de expertos de primer nivel en servicios de campo, asistencia técnica y piezas de repuesto certificadas.
Más información ›