Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12442
          Assay Name: hsa-miR-181b-5p
          miRBase Accession Number: MI0000270
          miRBase Version: v22.1
          Chromosome Location: Chr.1: 198858873 - 198858982 [-] on Build GRCh38
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Species: Human, Rat, Astatotilapia burtoni, Goat, Horse, Monodelphis domestica, Ophiophagus hannah, Pan troglodytes, Pongo pygmaeus, Tupaia chinensis
          Product Type: mirVana® miRNA mimic
          Assay ID MC12442

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000007, mir-181

          Mature miRNA Sequence:

          AACAUUCAUUGCUGUCGGUGGGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-181b-5p MIMAT0000257
          Rat rno-miR-181b-5p MIMAT0000859
          Astatotilapia burtoni abu-miR-181c MIMAT0042253
          Goat chi-miR-181b-5p MIMAT0036001
          Horse eca-miR-181b MIMAT0013179
          Monodelphis domestica mdo-miR-181b-5p MIMAT0004155
          View More Rows
          Ophiophagus hannah oha-miR-181b-5p MIMAT0036762
          Pan troglodytes ptr-miR-181b MIMAT0002625
          Pongo pygmaeus ppy-miR-181b MIMAT0002628
          Tupaia chinensis tch-miR-181b-5p MIMAT0036543

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-181b-1
          Stem-loop Accession # MI0000270
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000257
          miRBase ID: hsa-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38

          Stem-loop ID hsa-mir-181b-2
          Stem-loop Accession # MI0000683
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location Chr. 9 - 124693710 - 124693798 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000257
          miRBase ID: hsa-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 1 - 198858873 - 198858982 [-] on Build GRCh38

          Rat

          Stem-loop ID rno-mir-181b-1
          Stem-loop Accession # MI0000926
          Stem-loop Sequence
          CCUGUGCAGAGAUGAUGUUUACAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUAGAAAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000859
          miRBase ID: rno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0

          Stem-loop ID rno-mir-181b-2
          Stem-loop Accession # MI0000927
          Stem-loop Sequence
          UGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAAUGUCAACCAACUCACUGGUCAAUGAAUGCAAACUGCGGGCCAAAAA
          Chromosome Location Chr. 3 - 23151455 - 23151542 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000859
          miRBase ID: rno-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: Chr. 13 - 54952903 - 54953012 [+] on Build Rnor_6.0

          Astatotilapia burtoni

          Stem-loop ID abu-mir-181c
          Stem-loop Accession # MI0033987
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUGCAGCAGAAUAACUCACUGAUCAAUGAAUGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042253
          miRBase ID: abu-miR-181c
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-181b
          Stem-loop Accession # MI0030650
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGACUUUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036001
          miRBase ID: chi-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-181b
          Stem-loop Accession # MI0012921
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCAAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013179
          miRBase ID: eca-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-181b-1
          Stem-loop Accession # MI0005344
          Stem-loop Sequence
          AAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUUAACUGUAUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCACUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-181b-2
          Stem-loop Accession # MI0023753
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUUUUGAUUCAAUCAACUCACUGAUCAAUGAAUGCAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID mdo-mir-181b-3
          Stem-loop Accession # MI0023796
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGUGAGAUCUGAGGAAAAACUCACCGAUGGAUGAAUGUCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004155
          miRBase ID: mdo-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-181b
          Stem-loop Accession # MI0031378
          Stem-loop Sequence
          AAUAAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUCGGGUUCGGUCAGCCCUCUGGCUAAUGAAUGCAAACUGCGGAUCAAACAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036762
          miRBase ID: oha-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-181b-1
          Stem-loop Accession # MI0002934
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUUUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002625
          miRBase ID: ptr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID ptr-mir-181b-2
          Stem-loop Accession # MI0008556
          Stem-loop Sequence
          UGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002625
          miRBase ID: ptr-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-181b
          Stem-loop Accession # MI0002936
          Stem-loop Sequence
          CCUGUGCAGAGAUUAUUGUUUAAAAGGUCACAAUCAACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCAACUGUGGCCCCGCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002628
          miRBase ID: ppy-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Stem-loop ID ppy-mir-181b-2
          Stem-loop Accession # MI0014845
          Stem-loop Sequence
          CUGAUGGCUGCACUCAACAUUCAUUGCUGUCGGUGGGUUUGAGUCUGAAUCAACUCACUGAUCGAUGAAUGCAAACUGCGGACCAAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002628
          miRBase ID: ppy-miR-181b
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-181b
          Stem-loop Accession # MI0031203
          Stem-loop Sequence
          AACAUUCAUUGCUGUCGGUGGGUUGAACUGUGUGGACAAGCUCACUGAACAAUGAAUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036543
          miRBase ID: tch-miR-181b-5p
          Mature miRNA Sequence: AACAUUCAUUGCUGUCGGUGGGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465090_pri
          TaqMan™ Pri-miRNA Assay : Rn03464621_pri
          TaqMan™ Pri-miRNA Assay : Hs03302963_pri
          TaqMan™ Pri-miRNA Assay : Hs03303356_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12442
          Ambion® Pre-miR™ miRNA Precursor : PM12442
          mirVana® miRNA inhibitor : MH12442
          TaqMan™ Advanced miRNA Assay : rno478583_mir
          TaqMan™ Advanced miRNA Assay : 478583_mir
          TaqMan™ Advanced miRNA Assay : mmu478583_mir


          miRBase Alias:

          Human: hsa-miR-181b (v17)
          Mouse: mmu-miR-181b-5p (v21)
          Mouse: mmu-miR-181b (v17)
          Rat: rno-miR-181b (v19)
          Ochrobactrum anthropi: oan-miR-181b (v18)
          Ochrobactrum anthropi: oan-miR-181b-5p (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-pc8md:80/100.66.77.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0