Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10070
          Assay Name: hsa-miR-150-5p
          miRBase Accession Number: MI0000479
          miRBase Version: v22.1
          Chromosome Location: Chr.19: 49500785 - 49500868 [-] on Build GRCh38
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Callithrix jacchus, Cricetulus griseus, Dasypus novemcinctus, Dog, Goat, Gorilla gorilla, Guinea pig, Horse, Pan troglodytes, Pig, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Sheep
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10070

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000197, mir-150

          Mature miRNA Sequence:

          UCUCCCAACCCUUGUACCAGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-150-5p MIMAT0000451
          Mouse mmu-miR-150-5p MIMAT0000160
          Rat rno-miR-150-5p MIMAT0000853
          Alligator mississippiensis ami-miR-150-5p MIMAT0038180
          Anolis carolinensis aca-miR-150-5p MIMAT0021784
          Callithrix jacchus cja-miR-150 MIMAT0039474
          View More Rows
          Cricetulus griseus cgr-miR-150 MIMAT0041150
          Dasypus novemcinctus dno-miR-150-5p MIMAT0047675
          Dog cfa-miR-150 MIMAT0006602
          Goat chi-miR-150 MIMAT0035980
          Gorilla gorilla ggo-miR-150 MIMAT0024217
          Guinea pig cpo-miR-150-5p MIMAT0047028
          Horse eca-miR-150 MIMAT0013011
          Pan troglodytes ptr-miR-150 MIMAT0008044
          Pig ssc-miR-150 MIMAT0025365
          Pongo pygmaeus ppy-miR-150 MIMAT0015774
          Python bivittatus pbv-miR-150-5p MIMAT0038968
          Rhesus monkey mml-miR-150-5p MIMAT0006211
          Sheep oar-miR-150 MIMAT0030034

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-150
          Stem-loop Accession # MI0000479
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location Chr. 19 - 49500785 - 49500868 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000451
          miRBase ID: hsa-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 19 - 49500785 - 49500868 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-150
          Stem-loop Accession # MI0000172
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACCCUGGUACAGGCCUGGGGGAUAGGG
          Chromosome Location Chr. 7 - 45121757 - 45121821 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000160
          miRBase ID: mmu-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 7 - 45121757 - 45121821 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-150
          Stem-loop Accession # MI0000920
          Stem-loop Sequence
          CUUCUCAAGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACUUGGGGAC
          Chromosome Location Chr. 1 - 101115974 - 101116058 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000853
          miRBase ID: rno-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: Chr. 1 - 101115974 - 101116058 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-150
          Stem-loop Accession # MI0029756
          Stem-loop Sequence
          CCUUCUCUGCCCCCCACUCUCUCCCAACCCUUGUACCAGUGUCAUGUUACUGGAACCCUGGUACAGAGGAUGGAUGAGAAGGAGGCGUGGGACCCCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038180
          miRBase ID: ami-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-150
          Stem-loop Accession # MI0018761
          Stem-loop Sequence
          AAACUCUCUUUCCCUCUCUCCCAACCCUUGUACCAGUGCCAACUGUAGUCCUCACUGGUACAGAGGAUGGAAGGGUGGGCCUGCAGAGAUGAGACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021784
          miRBase ID: aca-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-150
          Stem-loop Accession # MI0031998
          Stem-loop Sequence
          UCCCCAUAGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGGCAGGGACCUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039474
          miRBase ID: cja-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-150
          Stem-loop Accession # MI0033079
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGCACUUCAGACCCUGGUACAGGCCUGGGGGGCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041150
          miRBase ID: cgr-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-150
          Stem-loop Accession # MI0039000
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGGCCUCAGACCCUGGUACAGGCCUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047675
          miRBase ID: dno-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-150
          Stem-loop Accession # MI0007998
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGUCCCUGGUACAGGCAUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006602
          miRBase ID: cfa-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-150
          Stem-loop Accession # MI0030638
          Stem-loop Sequence
          UCUCUUCUCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGUGUGUCUUGGUCCCUGGUACAGGUACGGGGAGGCAGGGACCUGGGGAUCCCAGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035980
          miRBase ID: chi-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-150
          Stem-loop Accession # MI0020764
          Stem-loop Sequence
          CUCCUCUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGACCCCGGCACCGGCAGGCCCCAAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0024217
          miRBase ID: ggo-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-150
          Stem-loop Accession # MI0038659
          Stem-loop Sequence
          UCUCCCAACCCUUGUACCAGUGCUGUGCCUCAGACGCUGGUACAGGCCUGGGGGGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047028
          miRBase ID: cpo-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-150
          Stem-loop Accession # MI0012762
          Stem-loop Sequence
          CCCUGUCUCCCAACCCUUGUACCAGUGCUGUGCCUCGGUCCCUGGUACAGGCAUGGGGGGCAGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013011
          miRBase ID: eca-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-150
          Stem-loop Accession # MI0008550
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008044
          miRBase ID: ptr-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-150-1
          Stem-loop Accession # MI0022133
          Stem-loop Sequence
          CUCGGCCCUAUCUCCCAACCCUUGUACCAGUGCGUGCCUCAGUCCCUGGUACAGGAAUGGGGGGCAGGGACCUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025365
          miRBase ID: ssc-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Stem-loop ID ssc-mir-150-2
          Stem-loop Accession # MI0022134
          Stem-loop Sequence
          CUCGGCCCUAUCUCCCAACCCUUGUACCAGUGCGUGCCUCAGUCCCUGGUACAGGAAUGGGGGGCAGGGACCUGGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025365
          miRBase ID: ssc-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-150
          Stem-loop Accession # MI0014840
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015774
          miRBase ID: ppy-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-150
          Stem-loop Accession # MI0030249
          Stem-loop Sequence
          ACUCUUCUCCUUUCUCUCCCAACCCUUGUACCAGUGUCAGCUGUUGCACCUACUGGUACAGAGGAUGGAAGGGAGAGUGGUUCAGAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038968
          miRBase ID: pbv-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-150
          Stem-loop Accession # MI0007641
          Stem-loop Sequence
          CUCCCCAUGGCCCUGUCUCCCAACCCUUGUACCAGUGCUGGGCUCAGACCCUGGUACAGGCCUGGGGGACAGGGACCUGGGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006211
          miRBase ID: mml-miR-150-5p
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-150
          Stem-loop Accession # MI0025255
          Stem-loop Sequence
          UCUCUCCUCACGGCCCUGUCUCCCAACCCUUGUACCAGUGUGUGUCUCGGUCCCUGGUACAGGUACGGGGAGGCAGGGACCUGGGGAUCCCAGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030034
          miRBase ID: oar-miR-150
          Mature miRNA Sequence: UCUCCCAACCCUUGUACCAGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03466044_pri
          TaqMan™ Pri-miRNA Assay : Hs03303271_pri
          TaqMan™ Pri-miRNA Assay : Mm03306366_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10070
          Ambion® Pre-miR™ miRNA Precursor : PM10070
          TaqMan™ MicroRNA Assay : 000473
          mirVana® miRNA mimic : MC10070
          TaqMan™ Advanced miRNA Assay : rno480947_mir
          TaqMan™ Advanced miRNA Assay : 477918_mir
          TaqMan™ Advanced miRNA Assay : mmu480947_mir


          miRBase Alias:

          Human: hsa-miR-150 (v17)
          Mouse: mmu-miR-150 (v17)
          Rat: rno-miR-150 (v18)
          Aeromonas caviae Ae398: aca-miR-150 (v18)
          Macaca mulatta: mml-miR-150 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline