Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10883
          Assay Name: hsa-miR-143-3p
          miRBase Accession Number: MI0000459
          miRBase Version: v22.1
          Chromosome Location: Chr.5: 149428918 - 149429023 [+] on Build GRCh38
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Species: Human, Mouse, Alligator mississippiensis, Anolis carolinensis, Astatotilapia burtoni, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Dog, Guinea pig, Horse, Ictalurus punctatus, Nomascus leucogenys, Oreochromis niloticus, Ornithorhynchus anatinus, Otolemur garnettii, Papio hamadryas, Pig, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Sheep, Xenopus laevis, Zebrafish
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10883

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000094, mir-143

          Mature miRNA Sequence:

          UGAGAUGAAGCACUGUAGCUC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-143-3p MIMAT0000435
          Mouse mmu-miR-143-3p MIMAT0000247
          Alligator mississippiensis ami-miR-143-3p MIMAT0038166
          Anolis carolinensis aca-miR-143-3p MIMAT0021770
          Astatotilapia burtoni abu-miR-143 MIMAT0042047
          Chrysemys picta cpi-miR-143-3p MIMAT0037769
          View More Rows
          Columba livia cli-miR-143-3p MIMAT0038532
          Cricetulus griseus cgr-miR-143 MIMAT0023772
          Dasypus novemcinctus dno-miR-143-3p MIMAT0047660
          Dog cfa-miR-143 MIMAT0006682
          Guinea pig cpo-miR-143-3p MIMAT0047011
          Horse eca-miR-143 MIMAT0013063
          Ictalurus punctatus ipu-miR-143 MIMAT0029420
          Nomascus leucogenys nle-miR-143 MIMAT0049427
          Oreochromis niloticus oni-miR-143 MIMAT0042647
          Ornithorhynchus anatinus oan-miR-143-3p MIMAT0007144
          Otolemur garnettii oga-miR-143 MIMAT0049667
          Papio hamadryas pha-miR-143 MIMAT0049514
          Pig ssc-miR-143-3p MIMAT0013879
          Pteropus alecto pal-miR-143-3p MIMAT0039781
          Python bivittatus pbv-miR-143-3p MIMAT0038956
          Rabbit ocu-miR-143-3p MIMAT0048248
          Rhesus monkey mml-miR-143-3p MIMAT0006201
          Salmo salar ssa-miR-143-3p MIMAT0032366
          Sheep oar-miR-143 MIMAT0030032
          Xenopus laevis xla-miR-143-3p MIMAT0046457
          Zebrafish dre-miR-143 MIMAT0001840

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-143
          Stem-loop Accession # MI0000459
          Stem-loop Sequence
          GCGCAGCGCCCUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGGAAGAGAGAAGUUGUUCUGCAGC
          Chromosome Location Chr. 5 - 149428918 - 149429023 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000435
          miRBase ID: hsa-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: Chr. 5 - 149428918 - 149429023 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-143
          Stem-loop Accession # MI0000257
          Stem-loop Sequence
          CCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGG
          Chromosome Location Chr. 18 - 61649196 - 61649258 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000247
          miRBase ID: mmu-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: Chr. 18 - 61649196 - 61649258 [-] on Build GRCm38

          Alligator mississippiensis

          Stem-loop ID ami-mir-143
          Stem-loop Accession # MI0029748
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038166
          miRBase ID: ami-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-143
          Stem-loop Accession # MI0018753
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021770
          miRBase ID: aca-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Astatotilapia burtoni

          Stem-loop ID abu-mir-143
          Stem-loop Accession # MI0033723
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGUAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042047
          miRBase ID: abu-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-143
          Stem-loop Accession # MI0029486
          Stem-loop Sequence
          AGCUCCGUCAAUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAACUGUGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037769
          miRBase ID: cpi-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-143
          Stem-loop Accession # MI0029978
          Stem-loop Sequence
          AUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAAUUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAACUGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038532
          miRBase ID: cli-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-143
          Stem-loop Accession # MI0020405
          Stem-loop Sequence
          CUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGGAAGGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023772
          miRBase ID: cgr-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-143
          Stem-loop Accession # MI0038992
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047660
          miRBase ID: dno-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-143
          Stem-loop Accession # MI0008092
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006682
          miRBase ID: cfa-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-143
          Stem-loop Accession # MI0038650
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047011
          miRBase ID: cpo-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-143
          Stem-loop Accession # MI0012807
          Stem-loop Sequence
          GCCCGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013063
          miRBase ID: eca-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Ictalurus punctatus

          Stem-loop ID ipu-mir-143
          Stem-loop Accession # MI0024509
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAACUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0029420
          miRBase ID: ipu-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-143
          Stem-loop Accession # MI0040169
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049427
          miRBase ID: nle-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Oreochromis niloticus

          Stem-loop ID oni-mir-143
          Stem-loop Accession # MI0034525
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGUAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0042647
          miRBase ID: oni-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-143
          Stem-loop Accession # MI0006871
          Stem-loop Sequence
          GAUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGGAGCUGUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007144
          miRBase ID: oan-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-143
          Stem-loop Accession # MI0040450
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049667
          miRBase ID: oga-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Papio hamadryas

          Stem-loop ID pha-mir-143
          Stem-loop Accession # MI0040279
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049514
          miRBase ID: pha-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-143
          Stem-loop Accession # MI0013098
          Stem-loop Sequence
          GUCCCCCAGCCGGAGGUGCAGUGCUGCAUCUCUGGUCAGCUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013879
          miRBase ID: ssc-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-143
          Stem-loop Accession # MI0032324
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039781
          miRBase ID: pal-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-143
          Stem-loop Accession # MI0030241
          Stem-loop Sequence
          UUCGAUGUCUCCCAGCCCAAGGUGCAGUGCUGCAUCUCUGGUCAGUUGUGAGUCUGAGAUGAAGCACUGUAGCUCAGGAAGGGAGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038956
          miRBase ID: pbv-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-143
          Stem-loop Accession # MI0039301
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048248
          miRBase ID: ocu-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-143
          Stem-loop Accession # MI0007632
          Stem-loop Sequence
          GCGCAGCGCCGUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCAGGAAGAGAGAAGUUGUUCUGCAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006201
          miRBase ID: mml-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-143
          Stem-loop Accession # MI0026503
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAAUUGGGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032366
          miRBase ID: ssa-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Sheep

          Stem-loop ID oar-mir-143
          Stem-loop Accession # MI0025253
          Stem-loop Sequence
          GCGUCCUGUCUCCCAGCCUGAGGUGCAGUGCUGCAUCUCUGGUCAGUUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAAGGGAGAAGUUGUUCUACAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030032
          miRBase ID: oar-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-mir-143
          Stem-loop Accession # MI0038254
          Stem-loop Sequence
          GGUGCAGUGCUGCAUCUCUGGUCAAUUGUGAGUCUGAGAUGAAGCACUGUAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046457
          miRBase ID: xla-miR-143-3p
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-143
          Stem-loop Accession # MI0002007
          Stem-loop Sequence
          GAUCUACAGUCGUCUGGCCCGCGGUGCAGUGCUGCAUCUCUGGUCAACUGGGAGUCUGAGAUGAAGCACUGUAGCUCGGGAGGACAACACUGUCAGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001840
          miRBase ID: dre-miR-143
          Mature miRNA Sequence: UGAGAUGAAGCACUGUAGCUC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03303166_pri
          TaqMan™ Pri-miRNA Assay : Mm03306564_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10883
          Ambion® Pre-miR™ miRNA Precursor : PM10883
          TaqMan™ MicroRNA Assay : 002249
          mirVana® miRNA mimic : MC10883
          TaqMan™ Advanced miRNA Assay : 477912_mir
          TaqMan™ Advanced miRNA Assay : mmu480935_mir


          miRBase Alias:

          Human: hsa-miR-143 (v17)
          Mouse: mmu-miR-143 (v17)
          Aeromonas caviae Ae398: aca-miR-143 (v18)
          Macaca mulatta: mml-miR-143 (v19)
          Ochrobactrum anthropi: oan-miR-143 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch