Search
Search

| Assay Name: | mmu-miR-669k-5p |
|---|---|
| miRBase Accession Number: | MI0006279 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UGUGCAUGUGUGUAUAGUUGUGUGC |
| Species: | Mouse |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000316, mir-467 |
Mature miRNA Sequence: | UGUGCAUGUGUGUAUAGUUGUGUGC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-669k-5p | MIMAT0017323 |