Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • mCherry Antibodies
          Now also available in recombinant format (Product # 740003T)

          Invitrogen

          mCherry Monoclonal Antibody (16D7)

          51 Published Figures
          266 References
          View all (42) mCherry antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite mCherry Monoclonal Antibody (16D7)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (9)
          • Published Figures (51)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 60

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          mCherry Antibody (M11217) in ICC/IF

          Immunofluorescent analysis of mCherry Tag was performed using 70% confluent mCherry-H3 transfected HEK-293 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 1 hour at room temperature. The cells were labeled with mCherry Monoclonal Antibody (16D7) (Product # M11217) at 5 µg/mL and Histone H3 Recombinant Polyclonal Antibody (Product # 711055) at 0.5 µg/mL in 0.1% BSA, incubated at 4°C overnight and then labeled with Goat anti-Rat IgG (H+L) C... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          mCherry Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Flow Cytometry (Flow)
          mCherry Antibody in Flow Cytometry (Flow)
          mCherry Antibody in Immunoprecipitation (IP)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          mCherry Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          mCherry Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          mCherry Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunocytochemistry (ICC/IF)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Immunohistochemistry (IHC)
          mCherry Antibody in Western Blot, Immunoprecipitation (WB, IP)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Western Blot (WB)
          mCherry Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          M11217

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1000
          View 31 publications 31 publications

          Immunohistochemistry (IHC)

          -
          View 147 publications 147 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 11 publications 11 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:100
          View 58 publications 58 publications

          Flow Cytometry (Flow)

          Assay-dependent
          View 6 publications 6 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 8 publications 8 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Tag

          Published species

          Tag

          Host/Isotype

          Rat / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          16D7

          Immunogen

          full-length protein mCherry

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Pacific Blue
          • Request custom conjugation

          Form

          Liquid

          Concentration

          2 mg/mL

          Amount

          200 µg

          Purification

          purified

          Storage buffer

          PBS

          Contains

          0.09% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2536611

          Target Information

          MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: mCherry

          View more View less

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-6b866bbc8c-mwps6:80/100.66.76.64:80.
          git-commit: 9d76681b284ad04e8ddcd97b79462898b32e8e5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.61.1-2026.10.99-1.0