Search
Search

| Assay Name: | mmu-miR-330 |
|---|---|
| miRBase Accession Number: | MI0000607 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | GCAAAGCACAGGGCCUGCAGAGA |
| Species: | Mouse, Rat, Cricetulus griseus |
| Product Type: | TaqMan™ MicroRNA Assay |
Gene Family ID: | MIPF0000200, mir-330 |
Mature miRNA Sequence: | GCAAAGCACAGGGCCUGCAGAGA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-330-3p | MIMAT0000569 |
| Rat | rno-miR-330-3p | MIMAT0000568 |
| Cricetulus griseus | cgr-miR-330 | MIMAT0041169 |