Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10732
          Assay Name: mmu-miR-330-3p
          miRBase Accession Number: MI0000607
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 19181465 - 19181562 [+] on Build GRCm38
          Mature miRNA Sequence: GCAAAGCACAGGGCCUGCAGAGA
          Species: Mouse, Rat, Cricetulus griseus
          Product Type: mirVana® miRNA mimic
          Assay ID MC10732

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000200, mir-330

          Mature miRNA Sequence:

          GCAAAGCACAGGGCCUGCAGAGA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-330-3p MIMAT0000569
          Rat rno-miR-330-3p MIMAT0000568
          Cricetulus griseus cgr-miR-330 MIMAT0041169

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-330
          Stem-loop Accession # MI0000607
          Stem-loop Sequence
          GACCCUUUGGCGAUCUCUGCCUCUCUGGGCCUGUGUCUUAGGCUCUUCAAGAUCCAACGAGCAAAGCACAGGGCCUGCAGAGAGGUAGCGCUCUGCUC
          Chromosome Location Chr. 7 - 19181465 - 19181562 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000569
          miRBase ID: mmu-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACAGGGCCUGCAGAGA
          Chromosome Location: Chr. 7 - 19181465 - 19181562 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-330
          Stem-loop Accession # MI0000606
          Stem-loop Sequence
          ACCCUUUGGCGAUCUCUGCCUCUCUGGGCCUGUGUCUUAGGCUCUUCAAGAUCUAACGAGCAAAGCACAGGGCCUGCAGAGAGGUAGCGCUCUGCUC
          Chromosome Location Chr. 1 - 80092681 - 80092777 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000568
          miRBase ID: rno-miR-330-3p
          Mature miRNA Sequence: GCAAAGCACAGGGCCUGCAGAGA
          Chromosome Location: Chr. 1 - 80092681 - 80092777 [+] on Build Rnor_6.0

          Cricetulus griseus

          Stem-loop ID cgr-mir-330
          Stem-loop Accession # MI0033099
          Stem-loop Sequence
          ACCCUUUGGCGAUCUCUGCCUCUCUGGGCCUGUGUCUUAGGCUCUUCAGGCUCUAACGAGCAAAGCACAGGGCCUGCAGAGAGGUAGCGCUCCGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0041169
          miRBase ID: cgr-miR-330
          Mature miRNA Sequence: GCAAAGCACAGGGCCUGCAGAGA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465914_pri
          TaqMan™ Pri-miRNA Assay : Mm03306963_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10732
          Ambion® Pre-miR™ miRNA Precursor : PM10732
          TaqMan™ MicroRNA Assay : 001062
          mirVana® miRNA inhibitor : MH10732
          TaqMan™ Advanced miRNA Assay : mmu481781_mir
          TaqMan™ Advanced miRNA Assay : rno481062_mir


          miRBase Alias:

          Mouse: mmu-miR-330* (v17)
          Rat: rno-miR-330* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline