Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC10120
          Assay Name: chi-miR-187
          miRBase Accession Number: MI0030658
          miRBase Version: v22.1
          Chromosome Location: -
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Species: Chicken, Fugu rubripes, Goat, Hippoglossus hippoglossus, Monodelphis domestica, Taeniopygia guttata, Tetraodon nigroviridis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC10120

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000078, mir-187

          Mature miRNA Sequence:

          UCGUGUCUUGUGUUGCAGCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Chicken gga-miR-187-3p MIMAT0001124
          Fugu rubripes fru-miR-187 MIMAT0002930
          Goat chi-miR-187 MIMAT0036012
          Hippoglossus hippoglossus hhi-miR-187 MIMAT0030356
          Monodelphis domestica mdo-miR-187-3p MIMAT0004120
          Taeniopygia guttata tgu-miR-187-3p MIMAT0014567
          View More Rows
          Tetraodon nigroviridis tni-miR-187 MIMAT0002931
          Zebrafish dre-miR-187 MIMAT0001274

          Stem-loop Details



          Chicken

          Stem-loop ID gga-mir-187
          Stem-loop Accession # MI0001193
          Stem-loop Sequence
          AAACUAUUGUGAGACCUCCGGCUACAACACAGGACAUGGGAGCUUUUCUGAACCCUCGUGUCUUGUGUUGCAGCCAGAGGGGCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001124
          miRBase ID: gga-miR-187-3p
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Fugu rubripes

          Stem-loop ID fru-mir-187
          Stem-loop Accession # MI0003246
          Stem-loop Sequence
          GUGGCCAUUUGGCCGGGCCAGGGGCUGCAACACAGGACAUGGGUCAUGCCUCUGCCCACCGCUCGUGUCUUGUGUUGCAGCCAGUGGAGCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002930
          miRBase ID: fru-miR-187
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-mir-187
          Stem-loop Accession # MI0030658
          Stem-loop Sequence
          GGCCGGGCUCCCAACGACACGGUGUGAGACCUCGGGCUACAACACGGGACCCGGGCGCUGCUCCGACCCCUCGUGUCUUGUGUUGCAGCCGGAGGGACGCAGGUCCGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036012
          miRBase ID: chi-miR-187
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Hippoglossus hippoglossus

          Stem-loop ID hhi-mir-187
          Stem-loop Accession # MI0025434
          Stem-loop Sequence
          GUGACCUCUCUGGCCGGGCCAAGGGCUGCAACACAGGACAUGGGUCCUGCUCUCCUCCCCGCUCGUGUCUUGUGUUGCAGCCAGUGGAGCUGCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0030356
          miRBase ID: hhi-miR-187
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-187
          Stem-loop Accession # MI0005309
          Stem-loop Sequence
          AUUGUGAGACCUCUGGCUACAACACAGGACACGGGAGCUUUUCUGAACCCUCGUGUCUUGUGUUGCAGCCAGAGGGGCACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004120
          miRBase ID: mdo-miR-187-3p
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-187
          Stem-loop Accession # MI0013788
          Stem-loop Sequence
          CUCCGGCUACAACACAGGACAUGGGAGCUUUUCUGAACCCUCGUGUCUUGUGUUGCAGCCAGAGGGGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014567
          miRBase ID: tgu-miR-187-3p
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Tetraodon nigroviridis

          Stem-loop ID tni-mir-187
          Stem-loop Accession # MI0003247
          Stem-loop Sequence
          GGCUGCAACACAGGACAUGGGUCUUGCCUCUGCCCGCCGCUCGUGUCUUGUGUUGCAGCCAGUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002931
          miRBase ID: tni-miR-187
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-mir-187
          Stem-loop Accession # MI0001370
          Stem-loop Sequence
          UGACCUGUGGCUGGGCCAGGGGCUGCAACACAGGACAUGGGAGCUGUCUCUCACUCCCGCUCGUGUCUUGUGUUGCAGCCAGUGGAACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0001274
          miRBase ID: dre-miR-187
          Mature miRNA Sequence: UCGUGUCUUGUGUUGCAGCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ MicroRNA Assay : 007816_mat
          mirVana® miRNA inhibitor : MH10120


          miRBase Alias:

          Gallus gallus: gga-miR-187 (v19)
          Monodelphis domestica: mdo-miR-187 (v19)
          Trabulsiella guamensis ATCC 49490: tgu-miR-187 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch