Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC11363
          Assay Name: rno-miR-363-3p
          miRBase Accession Number: MI0003553
          miRBase Version: v22.1
          Chromosome Location: Chr.X: 140117098 - 140117184 [-] on Build Rnor_6.0
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Species: Rat, Anolis carolinensis, Columba livia, Ornithorhynchus anatinus, Taeniopygia guttata, Xenopus tropicalis
          Product Type: mirVana® miRNA mimic
          Assay ID MC11363

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000138, mir-363

          Mature miRNA Sequence:

          AAUUGCACGGUAUCCAUCUGU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Rat rno-miR-363-3p MIMAT0003210
          Anolis carolinensis aca-miR-363-3p MIMAT0021946
          Columba livia cli-miR-363-3p MIMAT0038623
          Ornithorhynchus anatinus oan-miR-363-3p MIMAT0006860
          Taeniopygia guttata tgu-miR-363-3p MIMAT0014501
          Xenopus tropicalis xtr-miR-363-3p MIMAT0003702

          Stem-loop Details



          Rat

          Stem-loop ID rno-mir-363
          Stem-loop Accession # MI0003553
          Stem-loop Sequence
          UUUUGCUGUUAUCGGGUGGAUCACGAUGCAAUUUUGAUUAGAGUAAUGGGAGAGAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGA
          Chromosome Location Chr. X - 140117098 - 140117184 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0003210
          miRBase ID: rno-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: Chr. X - 140117098 - 140117184 [-] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-363
          Stem-loop Accession # MI0018860
          Stem-loop Sequence
          CUUGUGUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGAUUAGCAGGACGAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGACCUUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021946
          miRBase ID: aca-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-363
          Stem-loop Accession # MI0030034
          Stem-loop Sequence
          UUUGUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUUUAGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAAGACCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038623
          miRBase ID: cli-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-363
          Stem-loop Accession # MI0006698
          Stem-loop Sequence
          GUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUUUAGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006860
          miRBase ID: oan-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-363
          Stem-loop Accession # MI0013690
          Stem-loop Sequence
          GUUUUGCUGUUGUCGGGUGGAUCACGAUGCAAUUUUGAUUAGUUUAGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAAGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014501
          miRBase ID: tgu-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-363
          Stem-loop Accession # MI0004955
          Stem-loop Sequence
          UUUUGUUGUUCGCGGGUGGAUCACGAUGCAAUUUUAUUUAGUUUGGUAGGAGAAAAAUUGCACGGUAUCCAUCUGUAAACCGCAGAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003702
          miRBase ID: xtr-miR-363-3p
          Mature miRNA Sequence: AAUUGCACGGUAUCCAUCUGU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465449_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11363
          Ambion® Pre-miR™ miRNA Precursor : PM11363
          TaqMan™ MicroRNA Assay : 001355
          mirVana® miRNA inhibitor : MH11363
          TaqMan™ Advanced miRNA Assay : rno481384_mir


          miRBase Alias:

          Rat: rno-miR-363 (v18)
          Aeromonas caviae Ae398: aca-miR-363 (v18)
          Ochrobactrum anthropi: oan-miR-363 (v18)
          Trabulsiella guamensis ATCC 49490: tgu-miR-363 (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch