Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12580
          Assay Name: hsa-let-7c-3p
          miRBase Accession Number: MI0000064
          miRBase Version: v22.1
          Chromosome Location: Chr.21: 16539828 - 16539911 [+] on Build GRCh38
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Chicken, Chrysemys picta, Columba livia, Cricetulus griseus, Dasypus novemcinctus, Goat, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Paralichthys olivaceus, Pteropus alecto, Python bivittatus, Rabbit, Rhesus monkey, Salmo salar, Taeniopygia guttata, Xenopus laevis, Zebrafish
          Product Type: mirVana® miRNA mimic
          Assay ID MC12580

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000002, let-7

          Mature miRNA Sequence:

          CUGUACAACCUUCUAGCUUUCC

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-let-7c-3p MIMAT0026472
          Mouse mmu-let-7c-1-3p MIMAT0004622
          Rat rno-let-7c-1-3p MIMAT0017087
          Alligator mississippiensis ami-let-7c-3p MIMAT0038006
          Anolis carolinensis aca-let-7c-1-3p MIMAT0021697
          Chicken gga-let-7c-3p MIMAT0026492
          View More Rows
          Chrysemys picta cpi-let-7c-3p MIMAT0037600
          Columba livia cli-let-7c-3p MIMAT0038380
          Cricetulus griseus cgr-let-7c MIMAT0023718
          Dasypus novemcinctus dno-let-7c-3p MIMAT0047479
          Goat chi-let-7c-3p MIMAT0035885
          Guinea pig cpo-let-7c-3p MIMAT0046823
          Ophiophagus hannah oha-let-7c-3-3p MIMAT0036633
          Ornithorhynchus anatinus oan-let-7c-3p MIMAT0028959
          Paralichthys olivaceus pol-let-7d-3p MIMAT0025419
          Pteropus alecto pal-let-7c-3p MIMAT0040141
          Python bivittatus pbv-let-7c-3p MIMAT0038799
          Rabbit ocu-let-7c-3p MIMAT0048081
          Rhesus monkey mml-let-7c-3p MIMAT0026791
          Salmo salar ssa-let-7c-3p MIMAT0032266
          Salmo salar ssa-let-7d-3p MIMAT0032704
          Taeniopygia guttata tgu-let-7c-3p MIMAT0026983
          Xenopus laevis xla-let-7c-3p MIMAT0046400
          Zebrafish dre-let-7c-3p MIMAT0031930
          Zebrafish dre-let-7d-3p MIMAT0031931

          Stem-loop Details



          Human

          Stem-loop ID hsa-let-7c
          Stem-loop Accession # MI0000064
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGC
          Chromosome Location Chr. 21 - 16539828 - 16539911 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0026472
          miRBase ID: hsa-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: Chr. 21 - 16539828 - 16539911 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-let-7c-1
          Stem-loop Accession # MI0000559
          Stem-loop Sequence
          UGUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACU
          Chromosome Location Chr. 16 - 77599657 - 77599750 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004622
          miRBase ID: mmu-let-7c-1-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: Chr. 16 - 77599657 - 77599750 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-let-7c-1
          Stem-loop Accession # MI0000830
          Stem-loop Sequence
          UGUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACU
          Chromosome Location Chr. 11 - 16052873 - 16052966 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0017087
          miRBase ID: rno-let-7c-1-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: Chr. 11 - 16052873 - 16052966 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-let-7c-1
          Stem-loop Accession # MI0029644
          Stem-loop Sequence
          UGUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038006
          miRBase ID: ami-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-let-7c-1
          Stem-loop Accession # MI0018703
          Stem-loop Sequence
          GUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021697
          miRBase ID: aca-let-7c-1-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Chicken

          Stem-loop ID gga-let-7c
          Stem-loop Accession # MI0001174
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026492
          miRBase ID: gga-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-let-7c-1
          Stem-loop Accession # MI0029376
          Stem-loop Sequence
          GUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037600
          miRBase ID: cpi-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-let-7c
          Stem-loop Accession # MI0029887
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038380
          miRBase ID: cli-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-let-7c-1
          Stem-loop Accession # MI0020370
          Stem-loop Sequence
          GGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023718
          miRBase ID: cgr-let-7c
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-let-7c-1
          Stem-loop Accession # MI0038889
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047479
          miRBase ID: dno-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Goat

          Stem-loop ID chi-let-7c
          Stem-loop Accession # MI0030581
          Stem-loop Sequence
          GAAGCUGUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGAGCCAUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035885
          miRBase ID: chi-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-let-7c
          Stem-loop Accession # MI0038544
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046823
          miRBase ID: cpo-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-let-7c-3
          Stem-loop Accession # MI0031304
          Stem-loop Sequence
          AUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCAGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036633
          miRBase ID: oha-let-7c-3-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-let-7c-1
          Stem-loop Accession # MI0024053
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUCACACCCCGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028959
          miRBase ID: oan-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID oan-let-7c-2
          Stem-loop Accession # MI0024054
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUCACACCCCGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0028959
          miRBase ID: oan-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Paralichthys olivaceus

          Stem-loop ID pol-let-7d
          Stem-loop Accession # MI0022192
          Stem-loop Sequence
          UGAGGUAGUUGGUUGUAUGGUUUCGCAUAAUAAACAGCACGGAGAUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0025419
          miRBase ID: pol-let-7d-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-let-7c
          Stem-loop Accession # MI0032514
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0040141
          miRBase ID: pal-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-let-7c-1
          Stem-loop Accession # MI0030134
          Stem-loop Sequence
          GUGUGCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAAUUACACCCAGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038799
          miRBase ID: pbv-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-let-7c
          Stem-loop Accession # MI0039209
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048081
          miRBase ID: ocu-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-let-7c
          Stem-loop Accession # MI0007574
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026791
          miRBase ID: mml-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-let-7c-1
          Stem-loop Accession # MI0026423
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAAUGACACCCUAGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032266
          miRBase ID: ssa-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-let-7d-1
          Stem-loop Accession # MI0026754
          Stem-loop Sequence
          UGAGGUAGUUGGUUGUAUGGUUUCACAUAAUAAACAGCCCGGAGAUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032704
          miRBase ID: ssa-let-7d-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID ssa-let-7d-2
          Stem-loop Accession # MI0026755
          Stem-loop Sequence
          UGAGGUAGUUGGUUGUAUGGUUUCGCAUAAUAAACAGUCCGGAGAUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032704
          miRBase ID: ssa-let-7d-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-let-7c-1
          Stem-loop Accession # MI0013726
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026983
          miRBase ID: tgu-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID tgu-let-7c-2
          Stem-loop Accession # MI0013727
          Stem-loop Sequence
          GCAUCCGGGUUGAGGUAGUAGGUUGUAUGGUUUAGAGUUACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCACACUUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026983
          miRBase ID: tgu-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Xenopus laevis

          Stem-loop ID xla-let-7c
          Stem-loop Accession # MI0038221
          Stem-loop Sequence
          UGAGGUAGUAGGUUGUAUGGUUUAGAAUGACACCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046400
          miRBase ID: xla-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Zebrafish

          Stem-loop ID dre-let-7c-1
          Stem-loop Accession # MI0001866
          Stem-loop Sequence
          CUGAGAGUGUGUGCAUCCAGGCUGAGGUAGUAGGUUGUAUGGUUUAGAAUUUUGCCCUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGCUCACAAGCCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031930
          miRBase ID: dre-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID dre-let-7c-2
          Stem-loop Accession # MI0001867
          Stem-loop Sequence
          GUGUGCAUCCAGGCUGAGGUAGUAGGUUGUAUGGUUUCGAAUGACACCAUGGGAGUUAACUGUACAACCUUCUAGCUUUCCUUGGAGUACAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031930
          miRBase ID: dre-let-7c-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID dre-let-7d-1
          Stem-loop Accession # MI0001868
          Stem-loop Sequence
          UGUGCGUUGCGGUGUGAGGUAGUUGGUUGUAUGGUUUUGCAUAAUAAACAGCCCGGAGUUAACUGUACAACCUUCUAGCUUUCCCUGCGGCUGCACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031931
          miRBase ID: dre-let-7d-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA

          Stem-loop ID dre-let-7d-2
          Stem-loop Accession # MI0001870
          Stem-loop Sequence
          CGCUGCAGGCUGAGGUAGUUGGUUGUAUGGUUUUGCAUCAUAAUCAGCCUGGAGUUAACUGUACAACCUUCUAGCUUUCCCUGCGGUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0031931
          miRBase ID: dre-let-7d-3p
          Mature miRNA Sequence: CUGUACAACCUUCUAGCUUUCC
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306764_pri
          TaqMan™ Pri-miRNA Assay : Hs03302555_pri
          TaqMan™ Pri-miRNA Assay : Rn03464982_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12580
          Ambion® Pre-miR™ miRNA Precursor : PM12580
          TaqMan™ MicroRNA Assay : 002479
          mirVana® miRNA inhibitor : MH12580
          TaqMan™ Advanced miRNA Assay : 479365_mir
          TaqMan™ Advanced miRNA Assay : rno479365_mir
          TaqMan™ Advanced miRNA Assay : mmu483000_mir


          miRBase Alias:

          Mouse: mmu-let-7c-1* (v17)
          Rat: rno-let-7c-1* (v18)
          Aeromonas caviae Ae398: aca-let-7c-1* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-l8rg4:80/100.66.76.64:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0