Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC12998
          Assay Name: hsa-miR-10a-3p
          miRBase Accession Number: MI0000266
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 48579838 - 48579947 [-] on Build GRCh38
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Species: Human, Mouse, Rat, Columba livia, Dasypus novemcinctus, Guinea pig, Pteropus alecto, Rabbit
          Product Type: mirVana® miRNA mimic
          Assay ID MC12998

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          CAAAUUCGUAUCUAGGGGAAUA

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10a-3p MIMAT0004555
          Mouse mmu-miR-10a-3p MIMAT0004659
          Rat rno-miR-10a-3p MIMAT0004709
          Columba livia cli-miR-10a-3p MIMAT0038405
          Dasypus novemcinctus dno-miR-10a-3p MIMAT0047498
          Guinea pig cpo-miR-10a-3p MIMAT0046846
          View More Rows
          Pteropus alecto pal-miR-10a-3p MIMAT0039827
          Rabbit ocu-miR-10a-3p MIMAT0048098

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10a
          Stem-loop Accession # MI0000266
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCU
          Chromosome Location Chr. 17 - 48579838 - 48579947 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004555
          miRBase ID: hsa-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: Chr. 17 - 48579838 - 48579947 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-10a
          Stem-loop Accession # MI0000685
          Stem-loop Sequence
          GACCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location Chr. 11 - 96317165 - 96317274 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0004659
          miRBase ID: mmu-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: Chr. 11 - 96317165 - 96317274 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-10a
          Stem-loop Accession # MI0000841
          Stem-loop Sequence
          GACCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location Chr. 10 - 84165950 - 84166059 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0004709
          miRBase ID: rno-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: Chr. 10 - 84165950 - 84166059 [+] on Build Rnor_6.0

          Columba livia

          Stem-loop ID cli-mir-10a
          Stem-loop Accession # MI0029903
          Stem-loop Sequence
          CUUCUAUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAGGAAGUUGGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038405
          miRBase ID: cli-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-10a
          Stem-loop Accession # MI0038903
          Stem-loop Sequence
          UACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047498
          miRBase ID: dno-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-10a
          Stem-loop Accession # MI0038559
          Stem-loop Sequence
          UACCCUGUAGGUCCGAAUUUGUGUAAGGAAUUUUAUGGUCACAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046846
          miRBase ID: cpo-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-10a
          Stem-loop Accession # MI0032349
          Stem-loop Sequence
          UACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039827
          miRBase ID: pal-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-10a
          Stem-loop Accession # MI0039219
          Stem-loop Sequence
          UACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048098
          miRBase ID: ocu-miR-10a-3p
          Mature miRNA Sequence: CAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302873_pri
          TaqMan™ Pri-miRNA Assay : Rn03464991_pri
          TaqMan™ Pri-miRNA Assay : Mm03307058_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12998
          Ambion® Pre-miR™ miRNA Precursor : PM12998
          TaqMan™ MicroRNA Assay : 002288
          mirVana® miRNA inhibitor : MH12998
          TaqMan™ Advanced miRNA Assay : 478624_mir
          TaqMan™ Advanced miRNA Assay : mmu482997_mir
          TaqMan™ Advanced miRNA Assay : rno481317_mir


          miRBase Alias:

          Human: hsa-miR-10a* (v17)
          Mouse: mmu-miR-10a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-nqctq:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0