Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MC20577
          Assay Name: mmu-miR-210-5p
          miRBase Accession Number: MI0000695
          miRBase Version: v22.1
          Chromosome Location: Chr.7: 141221384 - 141221493 [-] on Build GRCm38
          Mature miRNA Sequence: AGCCACUGCCCACCGCACACUG
          Species: Mouse, Cricetulus griseus, Eptesicus fuscus, Guinea pig
          Product Type: mirVana® miRNA mimic
          Assay ID MC20577

          Purification
          Size
          Availability Made To Order
          Catalog # 4464066
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000086, mir-210

          Mature miRNA Sequence:

          AGCCACUGCCCACCGCACACUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-210-5p MIMAT0017052
          Cricetulus griseus cgr-miR-210-5p MIMAT0023848
          Eptesicus fuscus efu-miR-210 MIMAT0035147
          Guinea pig cpo-miR-210-5p MIMAT0047109

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-210
          Stem-loop Accession # MI0000695
          Stem-loop Sequence
          CCGGGGCAGUCCCUCCAGGCUCAGGACAGCCACUGCCCACCGCACACUGCGUUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCCCUGGGCAGCGCGAACC
          Chromosome Location Chr. 7 - 141221384 - 141221493 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0017052
          miRBase ID: mmu-miR-210-5p
          Mature miRNA Sequence: AGCCACUGCCCACCGCACACUG
          Chromosome Location: Chr. 7 - 141221384 - 141221493 [-] on Build GRCm38

          Cricetulus griseus

          Stem-loop ID cgr-mir-210
          Stem-loop Accession # MI0020458
          Stem-loop Sequence
          GGCUCAGGACAGCCACUGCCCACCGCACACUGCGUUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGUCCCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023848
          miRBase ID: cgr-miR-210-5p
          Mature miRNA Sequence: AGCCACUGCCCACCGCACACUG
          Chromosome Location: NA

          Eptesicus fuscus

          Stem-loop ID efu-mir-210
          Stem-loop Accession # MI0028852
          Stem-loop Sequence
          GGCGCAGGGCAGCCACUGCCCACCGCACACUGCGCUGCUCCGGACCCACUGUGCGUGUGACAGCGGCUGAUCUGCUCCUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0035147
          miRBase ID: efu-miR-210
          Mature miRNA Sequence: AGCCACUGCCCACCGCACACUG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-210
          Stem-loop Accession # MI0038702
          Stem-loop Sequence
          GCGCAGGGCAGCCACUGCCCACCGCACACUGCGCUGCUCCCGACCCACUGUGCGUGUGACAGCGGCUGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047109
          miRBase ID: cpo-miR-210-5p
          Mature miRNA Sequence: AGCCACUGCCCACCGCACACUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307104_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM20577
          Ambion® Pre-miR™ miRNA Precursor : PM20577
          TaqMan™ MicroRNA Assay : 462444_mat
          mirVana® miRNA inhibitor : MH20577
          TaqMan™ Advanced miRNA Assay : mmu481649_mir


          miRBase Alias:

          Mouse: mmu-miR-210* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch