Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10152
          Assay Name: hsa-miR-203a-3p
          miRBase Accession Number: MI0000283
          miRBase Version: v22.1
          Chromosome Location: Chr.14: 104117405 - 104117514 [+] on Build GRCh38
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Species: Human, Mouse, Rat, Callithrix jacchus, Dog, Guinea pig, Pan troglodytes, Pongo pygmaeus, Rhesus monkey
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10152

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000108, mir-203

          Mature miRNA Sequence:

          GUGAAAUGUUUAGGACCACUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-203a-3p MIMAT0000264
          Mouse mmu-miR-203-3p MIMAT0000236
          Rat rno-miR-203a-3p MIMAT0000876
          Callithrix jacchus cja-miR-203 MIMAT0039415
          Dog cfa-miR-203 MIMAT0009866
          Guinea pig cpo-miR-203-3p MIMAT0047098
          View More Rows
          Pan troglodytes ptr-miR-203 MIMAT0008065
          Pongo pygmaeus ppy-miR-203 MIMAT0015797
          Rhesus monkey mml-miR-203 MIMAT0006234

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-203a
          Stem-loop Accession # MI0000283
          Stem-loop Sequence
          GUGUUGGGGACUCGCGCGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGACCCGGCGGGCGCGGCGACAGCGA
          Chromosome Location Chr. 14 - 104117405 - 104117514 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000264
          miRBase ID: hsa-miR-203a-3p
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: Chr. 14 - 104117405 - 104117514 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-203
          Stem-loop Accession # MI0000246
          Stem-loop Sequence
          GCCUGGUCCAGUGGUUCUUGACAGUUCAACAGUUCUGUAGCACAAUUGUGAAAUGUUUAGGACCACUAGACCCGGC
          Chromosome Location Chr. 12 - 112130880 - 112130955 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000236
          miRBase ID: mmu-miR-203-3p
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: Chr. 12 - 112130880 - 112130955 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-203a
          Stem-loop Accession # MI0000945
          Stem-loop Sequence
          GCGCGCCUGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGACCCGGCGCGCACGGCAGCGGCGA
          Chromosome Location Chr. 6 - 136704961 - 136705057 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000876
          miRBase ID: rno-miR-203a-3p
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: Chr. 6 - 136704961 - 136705057 [+] on Build Rnor_6.0

          Callithrix jacchus

          Stem-loop ID cja-mir-203
          Stem-loop Accession # MI0031935
          Stem-loop Sequence
          CGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUGGCGCAAUUGUGAAAUGUUUAGGACCACUAGACCCGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039415
          miRBase ID: cja-miR-203
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA

          Dog

          Stem-loop ID cfa-mir-203
          Stem-loop Accession # MI0010363
          Stem-loop Sequence
          CCCGGCGCGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGCCCCGGCGGGCCCGGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009866
          miRBase ID: cfa-miR-203
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-203
          Stem-loop Accession # MI0038696
          Stem-loop Sequence
          AGUGGUUCUUAACAGUUCAACAGUUCUGUAGGGCAAUUGUGAAAUGUUUAGGACCACUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047098
          miRBase ID: cpo-miR-203-3p
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-203
          Stem-loop Accession # MI0008575
          Stem-loop Sequence
          UGUUGGGGACUCGCGCGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGACUCGGCGGGCGCGGCGACAGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0008065
          miRBase ID: ptr-miR-203
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-203
          Stem-loop Accession # MI0014862
          Stem-loop Sequence
          GUGUUGGGGACUCGCGCGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGACCCGGCGGGCACGGCGACAGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0015797
          miRBase ID: ppy-miR-203
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-203
          Stem-loop Accession # MI0007664
          Stem-loop Sequence
          GUGCUGGGGACUCGCGCGCUGGGUCCAGUGGUUCUUAACAGUUCAACAGUUCUGUAGCGCAAUUGUGAAAUGUUUAGGACCACUAGACCCGGCGGGCACGGCGACAGCGA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006234
          miRBase ID: mml-miR-203
          Mature miRNA Sequence: GUGAAAUGUUUAGGACCACUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Rn03465357_pri
          TaqMan™ Pri-miRNA Assay : Hs03302931_pri
          TaqMan™ Pri-miRNA Assay : Mm03306530_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10152
          Ambion® Pre-miR™ miRNA Precursor : PM10152
          TaqMan™ MicroRNA Assay : 000507
          mirVana® miRNA mimic : MC10152
          TaqMan™ Advanced miRNA Assay : 478316_mir
          TaqMan™ Advanced miRNA Assay : mmu481640_mir
          TaqMan™ Advanced miRNA Assay : rno478316_mir


          miRBase Alias:

          Human: hsa-miR-203 (v18)
          Human: hsa-miR-203a (v20)
          Mouse: mmu-miR-203 (v17)
          Rat: rno-miR-203 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch