Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH10787
          Assay Name: hsa-miR-10a-5p
          miRBase Accession Number: MI0000266
          miRBase Version: v22.1
          Chromosome Location: Chr.17: 48579838 - 48579947 [-] on Build GRCh38
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Ateles geoffroyi, Bovine, Branchiostoma floridae, Columba livia, Dasypus novemcinctus, Gorilla gorilla, Horse, Monodelphis domestica, Ophiophagus hannah, Pan paniscus, Pan troglodytes, Pongo pygmaeus, Python bivittatus, Rhesus monkey, Saccoglossus kowalevskii, Saguinus labiatus, Xenopus tropicalis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH10787

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000033, mir-10

          Mature miRNA Sequence:

          UACCCUGUAGAUCCGAAUUUGUG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-10a-5p MIMAT0000253
          Mouse mmu-miR-10a-5p MIMAT0000648
          Rat rno-miR-10a-5p MIMAT0000782
          Alligator mississippiensis ami-miR-10a-5p MIMAT0038030
          Anolis carolinensis aca-miR-10a-5p MIMAT0021717
          Ateles geoffroyi age-miR-10a MIMAT0002489
          View More Rows
          Bovine bta-miR-10a MIMAT0003786
          Branchiostoma floridae bfl-miR-10a-5p MIMAT0009466
          Columba livia cli-miR-10a-5p MIMAT0038404
          Dasypus novemcinctus dno-miR-10a-5p MIMAT0047497
          Gorilla gorilla ggo-miR-10a MIMAT0002486
          Horse eca-miR-10a MIMAT0013019
          Monodelphis domestica mdo-miR-10a-5p MIMAT0004089
          Ophiophagus hannah oha-miR-10a-5p MIMAT0036660
          Pan paniscus ppa-miR-10a MIMAT0002490
          Pan troglodytes ptr-miR-10a MIMAT0007944
          Pongo pygmaeus ppy-miR-10a MIMAT0002487
          Python bivittatus pbv-miR-10a-5p MIMAT0038824
          Rhesus monkey mml-miR-10a-5p MIMAT0006161
          Saccoglossus kowalevskii sko-miR-10 MIMAT0009611
          Saguinus labiatus sla-miR-10a MIMAT0002488
          Xenopus tropicalis xtr-miR-10a MIMAT0003557

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-10a
          Stem-loop Accession # MI0000266
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCU
          Chromosome Location Chr. 17 - 48579838 - 48579947 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000253
          miRBase ID: hsa-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: Chr. 17 - 48579838 - 48579947 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-10a
          Stem-loop Accession # MI0000685
          Stem-loop Sequence
          GACCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location Chr. 11 - 96317165 - 96317274 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000648
          miRBase ID: mmu-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: Chr. 11 - 96317165 - 96317274 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-10a
          Stem-loop Accession # MI0000841
          Stem-loop Sequence
          GACCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location Chr. 10 - 84165950 - 84166059 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000782
          miRBase ID: rno-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: Chr. 10 - 84165950 - 84166059 [+] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-10a
          Stem-loop Accession # MI0029663
          Stem-loop Sequence
          GUCUUCUAUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAGGAAGUUGCGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038030
          miRBase ID: ami-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-10a
          Stem-loop Accession # MI0018716
          Stem-loop Sequence
          GUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAAGAAACUGCGUCACAAAUUCGCGUCUAGGGGAAUAUGUAGUAGACACAAAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021717
          miRBase ID: aca-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Ateles geoffroyi

          Stem-loop ID age-mir-10a
          Stem-loop Accession # MI0002791
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002489
          miRBase ID: age-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Bovine

          Stem-loop ID bta-mir-10a
          Stem-loop Accession # MI0005007
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGAUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003786
          miRBase ID: bta-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Branchiostoma floridae

          Stem-loop ID bfl-mir-10a
          Stem-loop Accession # MI0010011
          Stem-loop Sequence
          CUAUAUGUACCCUGUAGAUCCGAAUUUGUGUGAGGUACCCAAGUCACAAAUUCGUUUCUGCAGGUAUAAUGUAGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009466
          miRBase ID: bfl-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-10a
          Stem-loop Accession # MI0029903
          Stem-loop Sequence
          CUUCUAUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAGGAAGUUGGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038404
          miRBase ID: cli-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-10a
          Stem-loop Accession # MI0038903
          Stem-loop Sequence
          UACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047497
          miRBase ID: dno-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Gorilla gorilla

          Stem-loop ID ggo-mir-10a
          Stem-loop Accession # MI0002788
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002486
          miRBase ID: ggo-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-10a
          Stem-loop Accession # MI0012768
          Stem-loop Sequence
          AAGAUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013019
          miRBase ID: eca-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Monodelphis domestica

          Stem-loop ID mdo-mir-10a
          Stem-loop Accession # MI0005273
          Stem-loop Sequence
          CUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0004089
          miRBase ID: mdo-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-10a
          Stem-loop Accession # MI0031319
          Stem-loop Sequence
          AAGGUCUCUCUGUCUUCUGUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAAGAAGCUGUCUCACAAAUUCGCAUCUAGGGGAAUAUGUAGUUGACACAAACCCUACGGAGCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036660
          miRBase ID: oha-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Pan paniscus

          Stem-loop ID ppa-mir-10a
          Stem-loop Accession # MI0002792
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002490
          miRBase ID: ppa-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Pan troglodytes

          Stem-loop ID ptr-mir-10a
          Stem-loop Accession # MI0008410
          Stem-loop Sequence
          AUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007944
          miRBase ID: ptr-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Pongo pygmaeus

          Stem-loop ID ppy-mir-10a
          Stem-loop Accession # MI0002789
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002487
          miRBase ID: ppy-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-10a
          Stem-loop Accession # MI0030153
          Stem-loop Sequence
          GUCUUCUGUAUGUACCCUGUAGAUCCGAAUUUGUGUAAAAGAAGCUGCCUCACAAAUUCGCAUCUAGGGGAAUAUGUAGUUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038824
          miRBase ID: pbv-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Rhesus monkey

          Stem-loop ID mml-mir-10a
          Stem-loop Accession # MI0007587
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006161
          miRBase ID: mml-miR-10a-5p
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Saccoglossus kowalevskii

          Stem-loop ID sko-mir-10
          Stem-loop Accession # MI0010149
          Stem-loop Sequence
          AUAUAUCUCCACCAACCUCUACCCUGUAGAUCCGAAUUUGUGGUACGUACAUUUUCACGAAUUCGUUUCUACCGGUGACAGGUAGCUGCGAGGACACAACA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0009611
          miRBase ID: sko-miR-10
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Saguinus labiatus

          Stem-loop ID sla-mir-10a
          Stem-loop Accession # MI0002790
          Stem-loop Sequence
          GAUCUGUCUGUCUUCUGUAUAUACCCUGUAGAUCCGAAUUUGUGUAAGGAAUUUUGUGGUCACAAAUUCGUAUCUAGGGGAAUAUGUAGUUGACAUAAACACUCCGCUCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0002488
          miRBase ID: sla-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA

          Xenopus tropicalis

          Stem-loop ID xtr-mir-10a
          Stem-loop Accession # MI0004796
          Stem-loop Sequence
          GAUUUGCCUGUCCUCUGUAUGUACCCUGUAGAUCCGAAUUUGUGUGAGCGCAAUCAUAUCACAAAUUCGUGUCUGGGGGGAUAUGCAGUUGACACAAACG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0003557
          miRBase ID: xtr-miR-10a
          Mature miRNA Sequence: UACCCUGUAGAUCCGAAUUUGUG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03307058_pri
          TaqMan™ Pri-miRNA Assay : Hs03302873_pri
          TaqMan™ Pri-miRNA Assay : Rn03464991_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM10787
          Ambion® Pre-miR™ miRNA Precursor : PM10787
          TaqMan™ MicroRNA Assay : 000387
          mirVana® miRNA mimic : MC10787
          TaqMan™ Advanced miRNA Assay : mmu479241_mir
          TaqMan™ Advanced miRNA Assay : 479241_mir
          TaqMan™ Advanced miRNA Assay : rno480893_mir


          miRBase Alias:

          Human: hsa-miR-10a (v17)
          Mouse: mmu-miR-10a (v17)
          Aeromonas caviae Ae398: aca-miR-10a (v18)
          Branchiostoma floridae: bfl-miR-10a (v18)
          Macaca mulatta: mml-miR-10a (v19)
          Monodelphis domestica: mdo-miR-10a (v19)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch