Search
Search

| Assay Name: | hsa-miR-519e-5p |
|---|---|
| miRBase Accession Number: | MI0003145 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | UUCUCCAAAAGGGAGCACUUUC |
| Species: | Human, Gorilla gorilla |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000020, mir-515 |
Mature miRNA Sequence: | UUCUCCAAAAGGGAGCACUUUC |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-519e-5p | MIMAT0002828 |
| Gorilla gorilla | ggo-miR-519e-5p | MIMAT0036407 |