Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH11679
          Assay Name: mmu-miR-133a-5p
          miRBase Accession Number: MI0000159
          miRBase Version: v22.1
          Chromosome Location: Chr.18: 10782909 - 10782976 [-] on Build GRCm38
          Mature miRNA Sequence: GCUGGUAAAAUGGAACCAAAU
          Species: Mouse
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH11679

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000029, mir-133

          Mature miRNA Sequence:

          GCUGGUAAAAUGGAACCAAAU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Mouse mmu-miR-133a-5p MIMAT0003473

          Stem-loop Details



          Mouse

          Stem-loop ID mmu-mir-133a-1
          Stem-loop Accession # MI0000159
          Stem-loop Sequence
          GCUAAAGCUGGUAAAAUGGAACCAAAUCGCCUCUUCAAUGGAUUUGGUCCCCUUCAACCAGCUGUAGC
          Chromosome Location Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003473
          miRBase ID: mmu-miR-133a-5p
          Mature miRNA Sequence: GCUGGUAAAAUGGAACCAAAU
          Chromosome Location: Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38

          Stem-loop ID mmu-mir-133a-2
          Stem-loop Accession # MI0000820
          Stem-loop Sequence
          AGAAGCCAAAUGCUUUGCUGAAGCUGGUAAAAUGGAACCAAAUCAGCUGUUGGAUGGAUUUGGUCCCCUUCAACCAGCUGUAGCUGCGCAUUGAUCACGCCGCA
          Chromosome Location Chr. 2 - 180398379 - 180398482 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0003473
          miRBase ID: mmu-miR-133a-5p
          Mature miRNA Sequence: GCUGGUAAAAUGGAACCAAAU
          Chromosome Location: Chr. 18 - 10782909 - 10782976 [-] on Build GRCm38


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Mm03306281_pri
          TaqMan™ Pri-miRNA Assay : Mm03307401_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM11679
          Ambion® Pre-miR™ miRNA Precursor : PM11679
          TaqMan™ MicroRNA Assay : 001637
          mirVana® miRNA mimic : MC11679
          TaqMan™ Advanced miRNA Assay : mmu481498_mir


          miRBase Alias:

          Mouse: mmu-miR-133a* (v17)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch