Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12412
          Assay Name: hsa-miR-17-5p
          miRBase Accession Number: MI0000071
          miRBase Version: v22.1
          Chromosome Location: Chr.13: 91350605 - 91350688 [+] on Build GRCh38
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Species: Human, Mouse, Rat, Alligator mississippiensis, Anolis carolinensis, Callithrix jacchus, Cricetulus griseus, Cyprinus carpio, Dasypus novemcinctus, Guinea pig, Horse, Microcebus murinus, Nomascus leucogenys, Ornithorhynchus anatinus, Otolemur garnettii, Pig, Pteropus alecto, Rabbit, Salmo salar, Taeniopygia guttata, Tupaia chinensis
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12412

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000001, mir-17

          Mature miRNA Sequence:

          CAAAGUGCUUACAGUGCAGGUAG

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-17-5p MIMAT0000070
          Mouse mmu-miR-17-5p MIMAT0000649
          Rat rno-miR-17-5p MIMAT0000786
          Alligator mississippiensis ami-miR-17-5p MIMAT0038047
          Anolis carolinensis aca-miR-17-5p MIMAT0021803
          Callithrix jacchus cja-miR-17 MIMAT0039391
          View More Rows
          Cricetulus griseus cgr-miR-17-5p MIMAT0023795
          Cyprinus carpio ccr-miR-17-5p MIMAT0026236
          Dasypus novemcinctus dno-miR-17-5p MIMAT0047509
          Guinea pig cpo-miR-17-5p MIMAT0046857
          Horse eca-miR-106a MIMAT0013194
          Horse eca-miR-17 MIMAT0013084
          Microcebus murinus mmr-miR-17 MIMAT0049198
          Nomascus leucogenys nle-miR-17 MIMAT0049455
          Ornithorhynchus anatinus oan-miR-17-5p MIMAT0007278
          Otolemur garnettii oga-miR-17 MIMAT0049648
          Pig ssc-miR-17-5p MIMAT0007755
          Pteropus alecto pal-miR-17-5p MIMAT0039959
          Rabbit ocu-miR-17-5p MIMAT0048109
          Salmo salar ssa-miR-17-5p MIMAT0032414
          Taeniopygia guttata tgu-miR-17a-5p MIMAT0017390
          Tupaia chinensis tch-miR-17-5p MIMAT0036516

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-17
          Stem-loop Accession # MI0000071
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGGUGAC
          Chromosome Location Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0000070
          miRBase ID: hsa-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. 13 - 91350605 - 91350688 [+] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-17
          Stem-loop Accession # MI0000687
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUGUGUGCAUCUACUGCAGUGAGGGCACUUGUAGCAUUAUGCUGAC
          Chromosome Location Chr. 14 - 115043671 - 115043754 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0000649
          miRBase ID: mmu-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. 14 - 115043671 - 115043754 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-17-1
          Stem-loop Accession # MI0000845
          Stem-loop Sequence
          GUCAGGAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGGUGUGUGCAUCUACUGCAGUGAAGGCACUUGUGGCAUUGUGCUGAC
          Chromosome Location Chr. 15 - 100179879 - 100179962 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000786
          miRBase ID: rno-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. X - 140117891 - 140117968 [-] on Build Rnor_6.0

          Stem-loop ID rno-mir-17-2
          Stem-loop Accession # MI0006131
          Stem-loop Sequence
          UUGGCCAUGUCAAAGUGCUUACAGUGCAGGUAGCUUCUUGAGAUCUACUGCACUGCAAGCACUUCUUACAUUACCAUG
          Chromosome Location Chr. X - 140117891 - 140117968 [-] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0000786
          miRBase ID: rno-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: Chr. X - 140117891 - 140117968 [-] on Build Rnor_6.0

          Alligator mississippiensis

          Stem-loop ID ami-mir-17
          Stem-loop Accession # MI0029672
          Stem-loop Sequence
          GUCAGAGUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUAUAGAACCUACUGCAGUGAAGGCACUUGUAGCAUUAUAGUGACAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038047
          miRBase ID: ami-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Anolis carolinensis

          Stem-loop ID aca-mir-17
          Stem-loop Accession # MI0018772
          Stem-loop Sequence
          AUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAAGUAGAACCUACUGCAGUGAAGGCACUUGUAGCAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021803
          miRBase ID: aca-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Callithrix jacchus

          Stem-loop ID cja-mir-17
          Stem-loop Accession # MI0031910
          Stem-loop Sequence
          AUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039391
          miRBase ID: cja-miR-17
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Cricetulus griseus

          Stem-loop ID cgr-mir-17
          Stem-loop Accession # MI0020419
          Stem-loop Sequence
          AGGAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAUGCACAUCUACUGCAGUGCAGGCACUUGUGGCAUUAUGGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0023795
          miRBase ID: cgr-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Cyprinus carpio

          Stem-loop ID ccr-mir-17
          Stem-loop Accession # MI0023342
          Stem-loop Sequence
          UCAAUGUAUUGUCAAAGUGCUUACAGUGCAGGUAGUAUUAUGGAAUAUCUACUGCAGUGGAGGCACUUCUAGCAAUAUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0026236
          miRBase ID: ccr-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Dasypus novemcinctus

          Stem-loop ID dno-mir-17
          Stem-loop Accession # MI0038909
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0047509
          miRBase ID: dno-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-17
          Stem-loop Accession # MI0038565
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046857
          miRBase ID: cpo-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Horse

          Stem-loop ID eca-mir-106a
          Stem-loop Accession # MI0012942
          Stem-loop Sequence
          CCUUGGCCAUGUCAAAGUGCUUACAGUGCAGGUAGCUUUCUGAGAUCUACUGCAAUGCAAGCACUUCUUACAUUACCAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013194
          miRBase ID: eca-miR-106a
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Stem-loop ID eca-mir-17
          Stem-loop Accession # MI0012831
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUAUGUGCAUCUACUGCAGUGAAGGCACUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0013084
          miRBase ID: eca-miR-17
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Microcebus murinus

          Stem-loop ID mmr-mir-17
          Stem-loop Accession # MI0039918
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049198
          miRBase ID: mmr-miR-17
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Nomascus leucogenys

          Stem-loop ID nle-mir-17
          Stem-loop Accession # MI0040207
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049455
          miRBase ID: nle-miR-17
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-17
          Stem-loop Accession # MI0006960
          Stem-loop Sequence
          GUCAGAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGGUAUGUAGAAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGUUGAC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007278
          miRBase ID: oan-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Otolemur garnettii

          Stem-loop ID oga-mir-17
          Stem-loop Accession # MI0040431
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0049648
          miRBase ID: oga-miR-17
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-17
          Stem-loop Accession # MI0008214
          Stem-loop Sequence
          GAAUAAUGUCAAAGUGCUUACAGUGCAGGUAGUGAUAAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGCAUUAUGG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0007755
          miRBase ID: ssc-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Pteropus alecto

          Stem-loop ID pal-mir-17
          Stem-loop Accession # MI0032421
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0039959
          miRBase ID: pal-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-17
          Stem-loop Accession # MI0039225
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048109
          miRBase ID: ocu-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Salmo salar

          Stem-loop ID ssa-mir-17-1
          Stem-loop Accession # MI0026535
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUCAUCUAAUAUCUACUGCAGUGAAGGCACUUUAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032414
          miRBase ID: ssa-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-17-2
          Stem-loop Accession # MI0026534
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUCUUCUAAUAUCUACUGCAGUGAAGGCACUUUAAGCA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032414
          miRBase ID: ssa-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Stem-loop ID ssa-mir-17-3
          Stem-loop Accession # MI0026533
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGUUAUUGAUGAUCUACUGCAGUGGAGGCACUUCUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0032414
          miRBase ID: ssa-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-17a
          Stem-loop Accession # MI0013784
          Stem-loop Sequence
          GUCAAAGUGCUUACAGUGCAGGUAGUGAUAUAUAGAACCUACUGCAGUGAAGGCACUUGUAGCAUUAUGUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0017390
          miRBase ID: tgu-miR-17a-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA

          Tupaia chinensis

          Stem-loop ID tch-mir-17
          Stem-loop Accession # MI0031174
          Stem-loop Sequence
          CAAAGUGCUUACAGUGCAGGUAGUGAUAUGUGCAUCUACUGCAGUGAAGGCACUUGUAGC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036516
          miRBase ID: tch-miR-17-5p
          Mature miRNA Sequence: CAAAGUGCUUACAGUGCAGGUAG
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03295901_pri
          TaqMan™ Pri-miRNA Assay : Rn03465733_pri
          TaqMan™ Pri-miRNA Assay : Rn03465594_pri
          TaqMan™ Pri-miRNA Assay : Mm04228426_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12412
          Ambion® Pre-miR™ miRNA Precursor : PM12412
          TaqMan™ MicroRNA Assay : 002308
          mirVana® miRNA mimic : MC12412
          TaqMan™ Advanced miRNA Assay : 478447_mir


          miRBase Alias:

          Human: hsa-miR-17 (v17)
          Mouse: mmu-miR-17 (v17)
          Aeromonas caviae Ae398: aca-miR-17 (v18)
          Ochrobactrum anthropi: oan-miR-17 (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7fb9f846c9-wx6nt:80/100.66.72.150:80.
          git-commit: 8207b5afa5c305bd40e0a4f0b9b53fd46f2a8829
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.59.1-2026.09.91-1.0