Search
Search

| Assay Name: | hsa-miR-155-3p |
|---|---|
| miRBase Accession Number: | MI0000681 |
| miRBase Version: | v22.1 |
| Mature miRNA Sequence: | CUCCUACAUAUUAGCAUUAACA |
| Species: | Human |
| Product Type: | mirVana® miRNA inhibitor |
Gene Family ID: | MIPF0000157, mir-155 |
Mature miRNA Sequence: | CUCCUACAUAUUAGCAUUAACA |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-155-3p | MIMAT0004658 |