Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › MH12646
          Assay Name: hsa-miR-30c-2-3p
          miRBase Accession Number: MI0000254
          miRBase Version: v22.1
          Chromosome Location: Chr.6: 71376960 - 71377031 [-] on Build GRCh38
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Species: Human, Mouse, Rat, Anolis carolinensis, Chrysemys picta, Columba livia, Guinea pig, Ophiophagus hannah, Ornithorhynchus anatinus, Pig, Python bivittatus, Rabbit, Taeniopygia guttata
          Product Type: mirVana® miRNA inhibitor
          Assay ID MH12646

          Purification
          Size
          Availability Made To Order
          Catalog # 4464084
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Family ID:

          MIPF0000005, mir-30

          Mature miRNA Sequence:

          CUGGGAGAAGGCUGUUUACUCU

          miRBase Details:



          Species miRBase ID miRBase Accession Number
          Human hsa-miR-30c-2-3p MIMAT0004550
          Mouse mmu-miR-30c-2-3p MIMAT0005438
          Rat rno-miR-30c-2-3p MIMAT0005442
          Anolis carolinensis aca-miR-30c-3p MIMAT0021925
          Chrysemys picta cpi-miR-30c-2-3p MIMAT0037680
          Columba livia cli-miR-30c-2-3p MIMAT0038455
          View More Rows
          Guinea pig cpo-miR-30c-2-3p MIMAT0046909
          Ophiophagus hannah oha-miR-30c-1-3p MIMAT0036894
          Ornithorhynchus anatinus oan-miR-30c-2-3p MIMAT0006912
          Pig ssc-miR-30c-3p MIMAT0022922
          Python bivittatus pbv-miR-30c-3p MIMAT0038874
          Rabbit ocu-miR-30c-2-3p MIMAT0048156
          Taeniopygia guttata tgu-miR-30c-3p MIMAT0014686

          Stem-loop Details



          Human

          Stem-loop ID hsa-mir-30c-2
          Stem-loop Accession # MI0000254
          Stem-loop Sequence
          AGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUUUCU
          Chromosome Location Chr. 6 - 71376960 - 71377031 [-] on Build GRCh38
          Mature MicroRNA miRBase Accession #: MIMAT0004550
          miRBase ID: hsa-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: Chr. 6 - 71376960 - 71377031 [-] on Build GRCh38

          Mouse

          Stem-loop ID mmu-mir-30c-2
          Stem-loop Accession # MI0000548
          Stem-loop Sequence
          GAGUGACAGAUAUUGUAAACAUCCUACACUCUCAGCUGUGAAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU
          Chromosome Location Chr. 1 - 23291701 - 23291784 [+] on Build GRCm38
          Mature MicroRNA miRBase Accession #: MIMAT0005438
          miRBase ID: mmu-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: Chr. 1 - 23291701 - 23291784 [+] on Build GRCm38

          Rat

          Stem-loop ID rno-mir-30c-2
          Stem-loop Accession # MI0000871
          Stem-loop Sequence
          GAGUGACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGAAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU
          Chromosome Location Chr. 9 - 29562376 - 29562459 [+] on Build Rnor_6.0
          Mature MicroRNA miRBase Accession #: MIMAT0005442
          miRBase ID: rno-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: Chr. 9 - 29562376 - 29562459 [+] on Build Rnor_6.0

          Anolis carolinensis

          Stem-loop ID aca-mir-30c-1
          Stem-loop Accession # MI0018849
          Stem-loop Sequence
          CCAAGUGACAGGUAAUGUAAACAUCCUACACUCUCAGCUGUGUAGAUAAGAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0021925
          miRBase ID: aca-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Chrysemys picta

          Stem-loop ID cpi-mir-30c-2
          Stem-loop Accession # MI0029427
          Stem-loop Sequence
          AGCUCUGAGUGGCAGGUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGGAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUGGAAAGCAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0037680
          miRBase ID: cpi-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Columba livia

          Stem-loop ID cli-mir-30c-2
          Stem-loop Accession # MI0029932
          Stem-loop Sequence
          UGACAGGUACUGUAAACAUCCUACACUCUCAGCUGUGGAAACUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCCCUGCC
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038455
          miRBase ID: cli-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Guinea pig

          Stem-loop ID cpo-mir-30c-2
          Stem-loop Accession # MI0038593
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGGACAGUAAGGAGCUGGGAGAAGGCUGUUUACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0046909
          miRBase ID: cpo-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Ophiophagus hannah

          Stem-loop ID oha-mir-30c-1
          Stem-loop Accession # MI0031453
          Stem-loop Sequence
          UGAUAGAGAGUGUAAACAUCCUACACUCUCAGCUGUGUGGAAAAGAUGAAAGCUGGGAGAAGGCUGUUUACUCUCUUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0036894
          miRBase ID: oha-miR-30c-1-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Ornithorhynchus anatinus

          Stem-loop ID oan-mir-30c-2
          Stem-loop Accession # MI0006728
          Stem-loop Sequence
          CUGUUCUAAAGAGCCCUGAGUGACAGGUACUGUAAACAUCCUACACUCUCAGCUGUAGAUGUGAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUGGAAAUCAGCCAAAGAGAACAG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0006912
          miRBase ID: oan-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Pig

          Stem-loop ID ssc-mir-30c-2
          Stem-loop Accession # MI0002461
          Stem-loop Sequence
          GACAGAUACUGUAAACAUCCUACACUCUCAGCUGUGGAAAGUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0022922
          miRBase ID: ssc-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Python bivittatus

          Stem-loop ID pbv-mir-30c-1
          Stem-loop Accession # MI0030185
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGUGGAAAAGAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCAUUGGAAA
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0038874
          miRBase ID: pbv-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Rabbit

          Stem-loop ID ocu-mir-30c-2
          Stem-loop Accession # MI0039250
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGGAAUGUAAGAAAGCUGGGAGAAGGCUGUUUACUCU
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0048156
          miRBase ID: ocu-miR-30c-2-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA

          Taeniopygia guttata

          Stem-loop ID tgu-mir-30c
          Stem-loop Accession # MI0013728
          Stem-loop Sequence
          UGUAAACAUCCUACACUCUCAGCUGUGGAAACUAAGAAAGCUGGGAGAAGGCUGUUUACUCUCUCUGCCUUG
          Chromosome Location -
          Mature MicroRNA miRBase Accession #: MIMAT0014686
          miRBase ID: tgu-miR-30c-3p
          Mature miRNA Sequence: CUGGGAGAAGGCUGUUUACUCU
          Chromosome Location: NA


          Back To Top

          More Information




          Related Products

          TaqMan™ Pri-miRNA Assay : Hs03302833_pri
          TaqMan™ Pri-miRNA Assay : Mm03306701_pri
          TaqMan™ Pri-miRNA Assay : Rn03465259_pri
          Ambion® Anti-miR™ miRNA Inhibitor : AM12646
          Ambion® Pre-miR™ miRNA Precursor : PM12646
          TaqMan™ MicroRNA Assay : 002110
          mirVana® miRNA mimic : MC12646
          TaqMan™ Advanced miRNA Assay : mmu479401_mir
          TaqMan™ Advanced miRNA Assay : rno479401_mir
          TaqMan™ Advanced miRNA Assay : 479401_mir


          miRBase Alias:

          Human: hsa-miR-30c-2* (v17)
          Mouse: mmu-miR-30c-2* (v17)
          Rat: rno-miR-30c-2* (v18)
          Aeromonas caviae Ae398: aca-miR-30c* (v18)
          Ochrobactrum anthropi: oan-miR-30c-3p (v19)
          Ochrobactrum anthropi: oan-miR-30c* (v18)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch