Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GPLD1 SIRNA Assays ›
          Gene Symbol:
          GPLD1
          Gene Name:
          glycosylphosphatidylinositol specific phospholipase D1
          Gene Aliases:
          GPIPLD, GPIPLDM, PIGPLD, PIGPLD1, PLD
          Chromosome Location:
          Chr.6: 24425832 - 24495163 on Build GRCh38
          UniGene ID:
          Hs.533291
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10171

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GPLD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001503.3 NP_001494.2 3, 4 351
          XM_011514509.2 XP_011512811.1 4, 5 421
          XM_017010753.1 XP_016866242.1 4, 5 423
          GenBank mRNA AJ308108.1 - 3, 4 351
          AK310692.1 - 3, 4 281
          AY007546.2 - 3, 4 245
          BC020748.1 - 3, 4 300
          BC093645.1 - 3, 4 270
          BC112001.1 - 3, 4 270
          L11701.1 - 3, 4 275
          L11702.1 - 3, 4 240

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase lipase phospholipase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          cell migration involved in sprouting angiogenesis
          chondrocyte differentiation
          complement receptor mediated signaling pathway
          GPI anchor release
          negative regulation of cell proliferation
          insulin receptor signaling pathway
          response to glucose
          positive regulation of endothelial cell migration
          positive regulation of alkaline phosphatase activity
          positive regulation of triglyceride biosynthetic process
          negative regulation of triglyceride catabolic process
          positive regulation of glucose metabolic process
          positive regulation of high-density lipoprotein particle clearance
          cellular response to insulin stimulus
          cellular response to drug
          hematopoietic stem cell migration
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          positive regulation of apoptotic process
          positive regulation of cytolysis
          phosphatidylcholine metabolic process
          positive regulation of membrane protein ectodomain proteolysis
          positive regulation of secretion
          transepithelial transport
          cellular response to calcium ion
          cellular response to cholesterol
          cellular response to triglyceride
          cellular response to pH
          hematopoietic stem cell migration to bone marrow
          regulation of cellular response to insulin stimulus
          glycosylphosphatidylinositol phospholipase D activity
          phospholipase D activity
          sodium channel regulator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0