Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GTF2H2C SIRNA Assays ›
          Gene Symbol:
          GTF2H2C GTF2H2 GTF2H2B GTF2H2C_2
          Gene Name:
          GTF2H2 family member C
          general transcription factor IIH subunit 2
          general transcription factor IIH subunit 2B (pseudogene)
          GTF2H2 family member C, copy 2
          Gene Aliases:
          GTF2H2C_2
          BTF2, BTF2P44, T-BTF2P44, TFIIH, p44
          -
          GTF2H2D
          Chromosome Location:
          Chr.5: 69560136 - 69592903 on Build GRCh38
          Chromosome Location:
          Chr.5: 71035124 - 71109278 on Build GRCh38
          Chromosome Location:
          Chr.5: 70415370 - 70450362 on Build GRCh38
          Chromosome Location:
          -
          UniGene ID:
          Hs.191356
          Hs.191356
          Hs.607501
          Hs.422901
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID 10173

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GTF2H2C
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098728.1 NP_001092198.1 10 791
          XM_005248588.4 XP_005248645.1 10 718
          XM_005248590.3 XP_005248647.1 10 656
          XM_005248592.3 XP_005248649.1 8 520
          XM_006714688.3 XP_006714751.1 10 709
          XM_011543615.1 XP_011541917.1 10 791
          XM_017009804.1 XP_016865293.1 8 574
          GenBank mRNA AK292361.1 - 10 770
          AK315282.1 - 10 791
          BC105980.1 - 10 649 4445643
          BC157894.1 - 10 721
          BC171860.1 - 11 783 9054514
          DQ786295.1 - 6 412
          Gene Symbol: GTF2H2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001515.3 NP_001506.1 10 788
          XM_017009403.1 XP_016864892.1 11 1397
          XM_017009404.1 XP_016864893.1 11 885
          XM_017009405.1 XP_016864894.1 11 740
          XM_017009406.1 XP_016864895.1 10 643
          GenBank mRNA AF078847.1 - 10 773
          AK296578.1 - 9 1010
          Z30094.1 - 9 578
          Gene Symbol: GTF2H2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_033417.1 9 755
          GenBank mRNA BC171919.1 - 9 684 9054573
          Gene Symbol: GTF2H2C_2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042490.3 NP_001035955.1 11 866
          GenBank mRNA BC064557.1 - 10 746 5549265
          BC065021.1 - 11 867 6047020
          BX647445.1 - 11 641

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein general transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleotide-excision repair
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          nucleotide-excision repair, DNA duplex unwinding
          G-protein coupled receptor internalization
          transcription-coupled nucleotide-excision repair
          nucleotide-excision repair, preincision complex stabilization
          nucleotide-excision repair, preincision complex assembly
          nucleotide-excision repair, DNA incision, 3'-to lesion
          nucleotide-excision repair, DNA incision, 5'-to lesion
          transcription initiation from RNA polymerase I promoter
          transcription elongation from RNA polymerase I promoter
          termination of RNA polymerase I transcription
          transcription from RNA polymerase II promoter
          transcription initiation from RNA polymerase II promoter
          transcription elongation from RNA polymerase II promoter
          7-methylguanosine mRNA capping
          translation
          protein phosphorylation
          response to UV
          nucleotide-excision repair, DNA incision
          global genome nucleotide-excision repair
          protein binding
          zinc ion binding
          nucleic acid binding
          transcription factor activity, sequence-specific DNA binding
          protein kinase activity
          DNA-dependent ATPase activity
          translation factor activity, RNA binding
          RNA polymerase II carboxy-terminal domain kinase activity
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0