Search
Search

Upk1b| Gene Symbol: Upk1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178924.4 | NP_849255.2 | 8 | 872 | ||
| GenBank mRNA | AK053004.1 | - | 8 | 873 | ||
| AK083101.1 | - | 8 | 872 | |||
| AK086926.1 | - | 8 | 871 | |||
| BC046604.1 | - | 8 | 850 | 6389837 | ||