Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP4A SIRNA Assays ›
          Gene Symbol:
          INPP4A
          Gene Name:
          inositol polyphosphate-4-phosphatase type I A
          Gene Aliases:
          INPP4, TVAS1
          Chromosome Location:
          Chr.2: 98444592 - 98594068 on Build GRCh38
          UniGene ID:
          Hs.469386
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10200

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INPP4A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001134224.1 NP_001127696.1 6, 7 782
          NM_001134225.1 NP_001127697.1 6, 7 782
          NM_001566.2 NP_001557.1 6, 7 782
          NM_004027.2 NP_004018.1 6, 7 782
          XM_006712499.3 XP_006712562.1 6, 7 777
          XM_006712500.3 XP_006712563.1 6, 7 777
          XM_006712501.3 XP_006712564.1 6, 7 777
          XM_006712502.3 XP_006712565.1 6, 7 777
          XM_006712503.3 XP_006712566.1 6, 7 777
          XM_006712504.3 XP_006712567.1 6, 7 777
          XM_006712505.3 XP_006712568.1 6, 7 777
          XM_006712506.3 XP_006712569.1 6, 7 777
          XM_006712507.3 XP_006712570.1 6, 7 777
          XM_006712508.3 XP_006712571.1 6, 7 777
          XM_011511122.2 XP_011509424.1 5, 6 727
          XM_011511123.2 XP_011509425.1 6, 7 777
          XM_011511124.2 XP_011509426.1 6, 7 777
          XM_011511125.2 XP_011509427.1 3, 4 659
          XM_017003993.1 XP_016859482.1 6, 7 618
          XM_017003994.1 XP_016859483.1 6, 7 777
          XM_017003995.1 XP_016859484.1 6, 7 777
          XM_017003996.1 XP_016859485.1 6, 7 777
          XM_017003997.1 XP_016859486.1 6, 7 836
          XM_017003998.1 XP_016859487.1 6, 7 836
          XM_017003999.1 XP_016859488.1 6, 7 835
          XM_017004000.1 XP_016859489.1 6, 7 777
          XM_017004001.1 XP_016859490.1 6, 7 777
          XM_017004002.1 XP_016859491.1 6, 7 695
          XM_017004003.1 XP_016859492.1 6, 7 777
          GenBank mRNA AF368319.1 - 4, 5 389
          BC028361.2 - 6, 7 782 4830663
          U26398.1 - 6, 7 683
          U96919.1 - 6, 7 683

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphatidylinositol biosynthetic process
          signal transduction
          dephosphorylation
          phosphatidylinositol-3-phosphate biosynthetic process
          inositol phosphate metabolic process
          protein binding
          phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
          inositol-1,3,4-trisphosphate 4-phosphatase activity
          phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity
          inositol-3,4-bisphosphate 4-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0