Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dbr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031403.3 | NP_113580.2 | 1 | 309 | ||
GenBank mRNA | AK050090.1 | - | 1 | 310 | ||
AK090351.1 | - | 1 | 285 | |||
AK139631.1 | - | 1 | 291 | |||
BC006661.1 | - | 1 | 304 | 3501390 |