Search
Search

Rab8a| Gene Symbol: Rab8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023126.2 | NP_075615.2 | 2 | 320 | ||
| GenBank mRNA | AB232600.1 | - | 2 | 161 | ||
| AK009259.1 | - | 2 | 171 | |||
| AK010862.1 | - | 2 | 171 | |||
| AK076048.1 | - | 2 | 199 | |||
| AK079306.1 | - | 187 | ||||
| AK080740.1 | - | 2 | 172 | |||
| AK147178.1 | - | 2 | 188 | |||
| AK165690.1 | - | 2 | 183 | |||
| AY974570.1 | - | 2 | 139 | |||
| BC019990.1 | - | 2 | 161 | 4161110 | ||
| BY279480.1 | - | 2 | 321 | |||
| S53270.1 | - | 2 | 178 | |||