Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABL2 SIRNA Assays ›
          Gene Symbol:
          ABL2
          Gene Name:
          ABL proto-oncogene 2, non-receptor tyrosine kinase
          Gene Aliases:
          ABLL, ARG
          Chromosome Location:
          Chr.1: 179099327 - 179229693 on Build GRCh38
          UniGene ID:
          Hs.159472
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103346

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABL2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001136000.2 NP_001129472.1 4 650
          NM_001136001.1 NP_001129473.1 3 874
          NM_001168236.1 NP_001161708.1 3 874
          NM_001168237.1 NP_001161709.1 4 937
          NM_001168238.1 NP_001161710.1 3 874
          NM_001168239.1 NP_001161711.1 3 587
          NM_005158.4 NP_005149.4 4 650
          NM_007314.3 NP_009298.1 4 937
          XM_005245088.2 XP_005245145.1 3 572
          XM_017001035.1 XP_016856524.1 3 3247
          GenBank mRNA AK225255.1 - 3 567
          AK309549.1 - 4 637
          AK311045.1 - 3 871
          BX538317.1 - 4 689
          FJ542283.1 - 3 542
          FJ542284.1 - 4 650
          FJ542285.1 - 3 587
          FJ542286.1 - 3 587
          M35296.1 - 4 854

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001107186, XM_006250050, XM_006250055, XM_006250049, XM_006250053, and others.
          This siRNA also targets the following transcripts in Mouse: NM_009595, NM_001136104, BC144897, BC144896, BC137771.

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          cell adhesion
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          positive regulation of cytosolic calcium ion concentration
          regulation of autophagy
          positive regulation of phospholipase C activity
          positive regulation of neuron projection development
          cell migration
          peptidyl-tyrosine phosphorylation
          actin cytoskeleton organization
          regulation of endocytosis
          regulation of cell adhesion
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          innate immune response
          positive regulation of oxidoreductase activity
          cellular response to retinoic acid
          regulation of cell motility
          regulation of actin cytoskeleton reorganization
          magnesium ion binding
          actin monomer binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          manganese ion binding
          actin filament binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0