Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PASK SIRNA Assays ›
Gene Symbol:
PASK
Gene Name:
PAS domain containing serine/threonine kinase
Gene Aliases:
PASKIN, STK37
Chromosome Location:
Chr.2: 241106099 - 241150347 on Build GRCh38
UniGene ID:
Hs.397891
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
103354
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PASK
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001252119.1
NP_001239048.1
10
1662
NM_001252120.1
NP_001239049.1
11
1815
NM_001252122.1
NP_001239051.1
10
1557
NM_001252124.1
NP_001239053.1
10
1662
NM_015148.3
NP_055963.2
10
1662
XM_005246991.1
XP_005247048.1
7
997
XM_011510826.1
XP_011509128.1
10
1617
XM_011510828.1
XP_011509130.1
11
1718
XM_011510829.1
XP_011509131.1
10
1659
XM_011510830.1
XP_011509132.1
10
1660
XM_011510831.2
XP_011509133.1
8
1824
XM_011510832.2
XP_011509134.1
8
1824
XM_011510833.1
XP_011509135.1
7
997
XM_011510834.1
XP_011509136.1
3
720
XM_011510835.1
XP_011509137.1
3
310
XM_017003634.1
XP_016859123.1
10
1906
XM_017003635.1
XP_016859124.1
11
1718
XM_017003636.1
XP_016859125.1
10
1674
GenBank mRNA
AF387103.1
-
10
1602
AK056516.1
-
10
1607
AK296945.1
-
7
1167
AK302417.1
-
10
1513
AK302561.1
-
7
997
AL117400.1
-
2
1961
BC020054.1
-
6
837
BC043495.1
-
4
419
BC050565.1
-
11
1621
4537404
BC063585.1
-
10
1605
4419998
CR749231.1
-
11
1816
D50925.2
-
10
1593
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
regulation of respiratory gaseous exchange
negative regulation of glycogen biosynthetic process
positive regulation of translation
protein autophosphorylation
regulation of glucagon secretion
regulation of energy homeostasis
protein serine/threonine kinase activity
protein binding
ATP binding
phosphatidylinositol binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline