Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERBB4 SIRNA Assays ›
          Gene Symbol:
          ERBB4
          Gene Name:
          erb-b2 receptor tyrosine kinase 4
          Gene Aliases:
          ALS19, HER4, p180erbB4
          Chromosome Location:
          Chr.2: 211375717 - 212539147 on Build GRCh38
          UniGene ID:
          Hs.390729
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103415

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ERBB4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042599.1 NP_001036064.1 6 735
          NM_005235.2 NP_005226.1 6 735
          XM_005246376.2 XP_005246433.1 6 1254
          XM_005246377.2 XP_005246434.1 6 1254
          XM_006712364.2 XP_006712427.1 6 1254
          XM_017003577.1 XP_016859066.1 7 1332
          XM_017003578.1 XP_016859067.1 7 1332
          XM_017003579.1 XP_016859068.1 7 1332
          XM_017003580.1 XP_016859069.1 7 1332
          XM_017003581.1 XP_016859070.1 7 1332
          XM_017003582.1 XP_016859071.1 2 367
          GenBank mRNA AK308606.1 - 6 691
          BC112199.1 - 6 676 8327667
          BC143741.1 - 6 664 9052255
          BC143747.1 - 6 664 9052261
          BC143749.1 - 6 664 9052263
          L07868.1 - 6 670

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          neural crest cell migration
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          heart development
          lactation
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          embryonic pattern specification
          regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of phosphatidylinositol 3-kinase signaling
          cell migration
          peptidyl-tyrosine phosphorylation
          central nervous system morphogenesis
          olfactory bulb interneuron differentiation
          regulation of cell migration
          ERBB2 signaling pathway
          positive regulation of tyrosine phosphorylation of Stat5 protein
          positive regulation of GTPase activity
          mitochondrial fragmentation involved in apoptotic process
          cell fate commitment
          positive regulation of transcription, DNA-templated
          protein autophosphorylation
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of cardiac muscle cell proliferation
          mammary gland epithelial cell differentiation
          mammary gland alveolus development
          cardiac muscle tissue regeneration
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of protein localization to cell surface
          regulation of cell motility
          positive regulation of STAT protein import into nucleus
          negative regulation of neuron migration
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          epidermal growth factor receptor binding
          protein binding
          ATP binding
          protein homodimerization activity
          transcription regulatory region DNA binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0