Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRC SIRNA Assays ›
          Gene Symbol:
          SRC
          Gene Name:
          SRC proto-oncogene, non-receptor tyrosine kinase
          Gene Aliases:
          ASV, SRC1, THC6, c-SRC, p60-Src
          Chromosome Location:
          Chr.20: 37344685 - 37405432 on Build GRCh38
          UniGene ID:
          Hs.195659
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103417

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRC
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005417.4 NP_005408.1 10 1402
          NM_198291.2 NP_938033.1 10 1327
          XM_011529013.2 XP_011527315.1 11 1671
          XM_017028024.1 XP_016883513.1 12 1695
          XM_017028025.1 XP_016883514.1 11 1576
          XM_017028026.1 XP_016883515.1 11 1415
          XM_017028027.1 XP_016883516.1 9 992
          GenBank mRNA AK024281.1 - 3 599
          BC011566.1 - 8 992 4548399
          BC051270.1 - 8 1055 4871614

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          cell cycle
          signal transduction
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          central nervous system development
          cell proliferation
          response to mechanical stimulus
          response to virus
          response to acidic pH
          regulation of epithelial cell migration
          positive regulation of epithelial cell migration
          positive regulation of platelet-derived growth factor receptor signaling pathway
          positive regulation of glucose metabolic process
          positive regulation of smooth muscle cell migration
          single organismal cell-cell adhesion
          peptidyl-serine phosphorylation
          peptidyl-tyrosine phosphorylation
          regulation of cell-cell adhesion
          platelet activation
          intracellular estrogen receptor signaling pathway
          forebrain development
          T cell costimulation
          protein destabilization
          response to nutrient levels
          positive regulation of protein autophosphorylation
          activation of protein kinase B activity
          negative regulation of telomere maintenance via telomerase
          negative regulation of protein homooligomerization
          cellular response to insulin stimulus
          regulation of intracellular estrogen receptor signaling pathway
          positive regulation of integrin activation
          adherens junction organization
          substrate adhesion-dependent cell spreading
          cellular response to reactive oxygen species
          intracellular signal transduction
          osteoclast development
          cellular response to platelet-derived growth factor stimulus
          peptidyl-tyrosine autophosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          ERBB2 signaling pathway
          regulation of cell proliferation
          response to drug
          response to hydrogen peroxide
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          regulation of vascular permeability
          stress fiber assembly
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of protein binding
          positive regulation of MAP kinase activity
          positive regulation of phosphatidylinositol 3-kinase activity
          transcytosis
          innate immune response
          regulation of bone resorption
          bone resorption
          positive regulation of cyclin-dependent protein serine/threonine kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of insulin receptor signaling pathway
          protein autophosphorylation
          platelet-derived growth factor receptor signaling pathway
          vascular endothelial growth factor receptor signaling pathway
          neurotrophin TRK receptor signaling pathway
          ephrin receptor signaling pathway
          oogenesis
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          progesterone receptor signaling pathway
          leukocyte migration
          positive regulation of small GTPase mediated signal transduction
          response to mineralocorticoid
          response to electrical stimulus
          regulation of cell cycle
          negative regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of mitochondrial depolarization
          negative regulation of telomerase activity
          uterus development
          branching involved in mammary gland duct morphogenesis
          regulation of cell projection assembly
          positive regulation of ERK1 and ERK2 cascade
          response to interleukin-1
          cellular response to lipopolysaccharide
          cellular response to peptide hormone stimulus
          cellular response to progesterone stimulus
          cellular response to fatty acid
          cellular response to hypoxia
          cellular response to fluid shear stress
          regulation of podosome assembly
          positive regulation of podosome assembly
          positive regulation of protein serine/threonine kinase activity
          angiotensin-activated signaling pathway involved in heart process
          positive regulation of canonical Wnt signaling pathway
          positive regulation of lamellipodium morphogenesis
          positive regulation of DNA biosynthetic process
          regulation of early endosome to late endosome transport
          negative regulation of anoikis
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway
          regulation of caveolin-mediated endocytosis
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          SH3/SH2 adaptor activity
          protein kinase C binding
          receptor binding
          insulin receptor binding
          integrin binding
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          enzyme binding
          kinase binding
          heme binding
          estrogen receptor binding
          SH2 domain binding
          ion channel binding
          ephrin receptor binding
          phosphoprotein binding
          hormone receptor binding
          growth factor receptor binding
          scaffold protein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gqkn5:80/100.66.76.113:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0