Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INSR SIRNA Assays ›
          Gene Symbol:
          INSR
          Gene Name:
          insulin receptor
          Gene Aliases:
          CD220, HHF5
          Chromosome Location:
          Chr.19: 7112255 - 7294405 on Build GRCh38
          UniGene ID:
          Hs.465744
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103467

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INSR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000208.3 NP_000199.2 13 2966
          NM_001079817.2 NP_001073285.1 12 2930
          XM_011527988.2 XP_011526290.2 13 3069
          XM_011527989.2 XP_011526291.2 12 3032
          GenBank mRNA AB208861.1 - 5 908
          BC117172.1 - 12 2552 40125723
          M10051.1 - 13 2693
          X02160.1 - 12 2567

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          negative regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          heart morphogenesis
          carbohydrate metabolic process
          regulation of transcription, DNA-templated
          G-protein coupled receptor signaling pathway
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          epidermis development
          male gonad development
          response to manganese ion
          regulation of hydrogen peroxide metabolic process
          positive regulation of glycoprotein biosynthetic process
          negative regulation of gene expression
          response to activity
          peptidyl-tyrosine phosphorylation
          transformation of host cell by virus
          signal transduction by protein phosphorylation
          male sex determination
          adrenal gland development
          positive regulation of cell migration
          exocrine pancreas development
          activation of protein kinase activity
          activation of protein kinase B activity
          response to estradiol
          negative regulation of transporter activity
          cellular response to insulin stimulus
          response to vitamin D
          response to testosterone
          response to tumor necrosis factor
          peptidyl-tyrosine autophosphorylation
          glucose homeostasis
          positive regulation of MAPK cascade
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          response to ethanol
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          regulation of embryonic development
          positive regulation of glucose import
          protein autophosphorylation
          positive regulation of developmental growth
          protein heterotetramerization
          response to glucocorticoid
          positive regulation of meiotic cell cycle
          positive regulation of protein kinase B signaling
          positive regulation of respiratory burst
          cellular response to growth factor stimulus
          regulation of female gonad development
          protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          insulin-activated receptor activity
          insulin-like growth factor receptor binding
          protein binding
          ATP binding
          GTP binding
          protein phosphatase binding
          lipoic acid binding
          insulin-like growth factor I binding
          insulin-like growth factor II binding
          protein complex binding
          3-phosphoinositide-dependent protein kinase binding
          phosphatidylinositol 3-kinase binding
          insulin binding
          insulin receptor substrate binding
          PTB domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0