Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTK2B SIRNA Assays ›
          Gene Symbol:
          PTK2B
          Gene Name:
          protein tyrosine kinase 2 beta
          Gene Aliases:
          CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK
          Chromosome Location:
          Chr.8: 27311482 - 27459391 on Build GRCh38
          UniGene ID:
          Hs.491322
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103485

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTK2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004103.4 NP_004094.3 16 1585
          NM_173174.2 NP_775266.1 20 2140
          NM_173175.2 NP_775267.1 15 1487
          NM_173176.2 NP_775268.1 15 1572
          XM_005273447.4 XP_005273504.1 15 1500
          XM_005273448.4 XP_005273505.1 15 1549
          XM_011544441.2 XP_011542743.1 15 1510
          XM_011544442.2 XP_011542744.1 15 1510
          XM_017013214.1 XP_016868703.1 16 1575
          XM_017013215.1 XP_016868704.1 15 1471
          XM_017013216.1 XP_016868705.1 15 1510
          GenBank mRNA AK128371.1 - 7 918
          U33284.1 - 15 1439

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          angiogenesis
          oocyte maturation
          response to hypoxia
          positive regulation of cell-matrix adhesion
          sprouting angiogenesis
          adaptive immune response
          marginal zone B cell differentiation
          protein complex assembly
          protein phosphorylation
          apoptotic process
          response to stress
          cellular defense response
          response to osmotic stress
          signal transduction
          cell surface receptor signaling pathway
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          integrin-mediated signaling pathway
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell shape
          response to mechanical stimulus
          response to hormone
          response to glucose
          response to lithium ion
          positive regulation of endothelial cell migration
          negative regulation of muscle cell apoptotic process
          regulation of cGMP-mediated signaling
          regulation of macrophage chemotaxis
          positive regulation of neuron projection development
          glial cell proliferation
          peptidyl-tyrosine phosphorylation
          regulation of cell adhesion
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of bone mineralization
          regulation of cGMP biosynthetic process
          positive regulation of actin filament polymerization
          neuron projection development
          regulation of inositol trisphosphate biosynthetic process
          tumor necrosis factor-mediated signaling pathway
          ionotropic glutamate receptor signaling pathway
          response to immobilization stress
          peptidyl-tyrosine autophosphorylation
          response to cocaine
          response to drug
          response to hydrogen peroxide
          activation of Janus kinase activity
          negative regulation of apoptotic process
          stress fiber assembly
          negative regulation of potassium ion transport
          positive regulation of JUN kinase activity
          negative regulation of neuron apoptotic process
          blood vessel endothelial cell migration
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          positive regulation of nitric oxide biosynthetic process
          bone resorption
          response to ethanol
          negative regulation of myeloid cell differentiation
          positive regulation of translation
          positive regulation of angiogenesis
          positive regulation of protein kinase activity
          positive regulation of JNK cascade
          protein autophosphorylation
          vascular endothelial growth factor receptor signaling pathway
          focal adhesion assembly
          positive regulation of peptidyl-tyrosine phosphorylation
          regulation of calcium-mediated signaling
          positive regulation of nitric-oxide synthase activity
          regulation of release of sequestered calcium ion into cytosol
          response to cAMP
          response to calcium ion
          positive regulation of synaptic transmission, glutamatergic
          long-term synaptic potentiation
          long term synaptic depression
          chemokine-mediated signaling pathway
          positive regulation of ERK1 and ERK2 cascade
          cellular response to retinoic acid
          cellular response to fluid shear stress
          activation of GTPase activity
          regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          regulation of establishment of cell polarity
          regulation of actin cytoskeleton reorganization
          regulation of N-methyl-D-aspartate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          positive regulation of B cell chemotaxis
          positive regulation of DNA biosynthetic process
          calmodulin-dependent protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          NMDA glutamate receptor activity
          receptor binding
          protein binding
          ATP binding
          protein complex binding
          3-phosphoinositide-dependent protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0