Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PASK SIRNA Assays ›
          Gene Symbol:
          PASK
          Gene Name:
          PAS domain containing serine/threonine kinase
          Gene Aliases:
          PASKIN, STK37
          Chromosome Location:
          Chr.2: 241106099 - 241150347 on Build GRCh38
          UniGene ID:
          Hs.397891
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103518

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PASK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252119.1 NP_001239048.1 12, 13 3198
          NM_001252120.1 NP_001239049.1 13, 14 3351
          NM_001252122.1 NP_001239051.1 12, 13 3093
          NM_001252124.1 NP_001239053.1 12, 13 3198
          NM_015148.3 NP_055963.2 12, 13 3198
          XM_005246991.1 XP_005247048.1 9, 10 2533
          XM_011510826.1 XP_011509128.1 12, 13 3153
          XM_011510828.1 XP_011509130.1 13, 14 3254
          XM_011510829.1 XP_011509131.1 12, 13 3195
          XM_011510830.1 XP_011509132.1 12, 13 3196
          XM_011510831.2 XP_011509133.1 10, 11 3360
          XM_011510832.2 XP_011509134.1 10, 11 3360
          XM_011510833.1 XP_011509135.1 9, 10 2533
          XM_011510834.1 XP_011509136.1 5, 6 2256
          XM_011510835.1 XP_011509137.1 5, 6 1846
          XM_017003634.1 XP_016859123.1 12, 13 3442
          XM_017003635.1 XP_016859124.1 13, 14 3254
          XM_017003636.1 XP_016859125.1 12, 13 3210
          GenBank mRNA AK302417.1 - 12, 13 3049
          AK302561.1 - 9, 10 2533
          AL117400.1 - 4, 5 3497
          BC043495.1 - 6, 7 1955
          BC050565.1 - 13, 14 3157 4537404
          BC063585.1 - 12, 13 3141 4419998
          CR749231.1 - 13, 14 3352
          D50925.2 - 12, 13 3129
          U79240.1 - 3, 4 472

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of respiratory gaseous exchange
          negative regulation of glycogen biosynthetic process
          positive regulation of translation
          protein autophosphorylation
          regulation of glucagon secretion
          regulation of energy homeostasis
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline