Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKAA2 SIRNA Assays ›
          Gene Symbol:
          PRKAA2
          Gene Name:
          protein kinase AMP-activated catalytic subunit alpha 2
          Gene Aliases:
          AMPK, AMPK2, AMPKa2, PRKAA
          Chromosome Location:
          Chr.1: 56645317 - 56715335 on Build GRCh38
          UniGene ID:
          Hs.437039
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103652

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKAA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006252.3 NP_006243.2 5, 6 638
          XM_017001692.1 XP_016857181.1 5, 6 2103
          XM_017001693.1 XP_016857182.1 5, 6 723
          XM_017001694.1 XP_016857183.1 6, 7 631
          GenBank mRNA AK314011.1 - 5, 6 638
          AL710110.1 - 1, 2 48
          BC069680.1 - 5, 6 610 7262537
          BC069740.1 - 5, 6 611 7262549
          BC069823.1 - 5, 6 611 7262561
          EF056019.1 - 5, 6 567
          FJ200485.1 - 3, 4 331
          FJ200486.1 - 4, 5 473
          HQ628625.1 - 4, 5 425
          U06454.1 - 5, 6 631

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein phosphorylation
          fatty acid biosynthetic process
          cholesterol biosynthetic process
          carnitine shuttle
          response to stress
          cell cycle arrest
          signal transduction
          lipid biosynthetic process
          positive regulation of autophagy
          response to muscle activity
          Wnt signaling pathway
          macroautophagy
          positive regulation of macroautophagy
          regulation of macroautophagy
          cellular response to nutrient levels
          negative regulation of TOR signaling
          histone-serine phosphorylation
          intracellular signal transduction
          cellular response to drug
          cellular response to glucose starvation
          regulation of fatty acid biosynthetic process
          glucose homeostasis
          regulation of circadian rhythm
          negative regulation of apoptotic process
          positive regulation of glycolytic process
          rhythmic process
          fatty acid homeostasis
          cellular response to prostaglandin E stimulus
          regulation of signal transduction by p53 class mediator
          regulation of energy homeostasis
          chromatin binding
          protein kinase activity
          protein serine/threonine kinase activity
          AMP-activated protein kinase activity
          protein serine/threonine/tyrosine kinase activity
          protein binding
          ATP binding
          histone serine kinase activity
          metal ion binding
          [hydroxymethylglutaryl-CoA reductase (NADPH)] kinase activity
          [acetyl-CoA carboxylase] kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0