Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKCD SIRNA Assays ›
          Gene Symbol:
          PRKCD
          Gene Name:
          protein kinase C delta
          Gene Aliases:
          ALPS3, CVID9, MAY1, PKCD, nPKC-delta
          Chromosome Location:
          Chr.3: 53161207 - 53192717 on Build GRCh38
          UniGene ID:
          Hs.155342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 103653

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKCD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001316327.1 NP_001303256.1 9 896
          NM_006254.3 NP_006245.2 9 1063
          NM_212539.1 NP_997704.1 8 951
          XM_006713257.3 XP_006713320.1 8 870
          XM_006713259.2 XP_006713322.1 10 1167
          XM_017006855.1 XP_016862344.1 8 748
          XM_017006856.1 XP_016862345.1 8 1322
          GenBank mRNA AK130150.1 - 4 254
          AK131548.1 - 1411
          AK294272.1 - 6 522
          AK313216.1 - 9 896
          BC043350.1 - 9 1082 5539909
          D10495.1 - 6 569
          DQ516383.1 - 1 41
          L07860.1 - 8 768
          L07861.1 - 8 768
          Z22521.1 - 8 1039

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          stimulatory C-type lectin receptor signaling pathway
          protein phosphorylation
          apoptotic process
          cellular component disassembly involved in execution phase of apoptosis
          cell cycle
          signal transduction
          activation of phospholipase C activity
          intrinsic apoptotic signaling pathway in response to oxidative stress
          regulation of receptor activity
          immunoglobulin mediated immune response
          histone phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          peptidyl-tyrosine phosphorylation
          termination of signal transduction
          platelet activation
          negative regulation of actin filament polymerization
          positive regulation of endodeoxyribonuclease activity
          negative regulation of protein binding
          activation of protein kinase activity
          interleukin-10 production
          interleukin-12 production
          positive regulation of superoxide anion generation
          regulation of actin cytoskeleton organization
          negative regulation of glial cell apoptotic process
          positive regulation of protein dephosphorylation
          Fc-gamma receptor signaling pathway involved in phagocytosis
          B cell proliferation
          neutrophil activation
          positive regulation of protein import into nucleus
          defense response to bacterium
          negative regulation of MAP kinase activity
          regulation of mRNA stability
          negative regulation of insulin receptor signaling pathway
          negative regulation of inflammatory response
          negative regulation of peptidyl-tyrosine phosphorylation
          protein stabilization
          negative regulation of filopodium assembly
          cell chemotaxis
          interferon-gamma-mediated signaling pathway
          cellular response to hydrogen peroxide
          cellular response to hydroperoxide
          negative regulation of platelet aggregation
          cellular senescence
          positive regulation of phospholipid scramblase activity
          cellular response to angiotensin
          positive regulation of ceramide biosynthetic process
          positive regulation of glucosylceramide catabolic process
          positive regulation of sphingomyelin catabolic process
          positive regulation of response to DNA damage stimulus
          positive regulation of apoptotic signaling pathway
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          enzyme activator activity
          enzyme binding
          kinase binding
          protein kinase binding
          insulin receptor substrate binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0