Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ARF1 SIRNA Assays ›
          Gene Symbol:
          ARF1
          Gene Name:
          ADP ribosylation factor 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 228082660 - 228099212 on Build GRCh38
          UniGene ID:
          Hs.286221
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 10426

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ARF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001024226.1 NP_001019397.1 5 1733
          NM_001024227.1 NP_001019398.1 5 1767
          NM_001024228.1 NP_001019399.1 5 1745
          NM_001658.3 NP_001649.1 5 1648
          XM_017001233.1 XP_016856722.1 4 1736
          GenBank mRNA AF052179.1 - 5 1580
          AF055002.1 - 5 1575
          AK000620.1 - 5 1648
          AK057838.1 - 5 1576
          AK057875.1 - 5 1586
          AK130737.1 - 5 1577
          AK296020.1 - 5 1496
          BC009247.2 - 5 1560 2963635
          BC010429.1 - 5 1573 3356859
          BC011358.1 - 5 1599 3463523
          BF337061.1 - 1 377
          M36340.1 - 5 1578
          M84326.1 - 5 1577

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008767870, XM_006246525, FQ219813, FQ213372, BC061552, and others.
          This siRNA also targets the following transcripts in Mouse: NM_007476, NM_001130408, BC031986, AK168686, AK170586, and others.

          Panther Classification:

          Molecular Function -

          G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of receptor internalization
          phosphatidylinositol biosynthetic process
          cellular copper ion homeostasis
          post-Golgi vesicle-mediated transport
          actin filament organization
          small GTPase mediated signal transduction
          protein transport
          antigen processing and presentation of exogenous peptide antigen via MHC class II
          regulation of Arp2/3 complex-mediated actin nucleation
          very-low-density lipoprotein particle assembly
          positive regulation of catalytic activity
          positive regulation of endocytosis
          positive regulation of calcium ion-dependent exocytosis
          regulation of defense response to virus by virus
          positive regulation of protein secretion
          Golgi to transport vesicle transport
          long term synaptic depression
          positive regulation of dendritic spine development
          synaptic vesicle budding
          dendritic spine organization
          lysosomal membrane organization
          positive regulation of sodium ion transmembrane transport
          positive regulation of late endosome to lysosome transport
          positive regulation of ER to Golgi vesicle-mediated transport
          regulation of phospholipid metabolic process
          magnesium ion binding
          GTPase activity
          receptor signaling protein activity
          protein binding
          GTP binding
          GDP binding
          poly(A) RNA binding
          phospholipase D activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0