Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1C SIRNA Assays ›
Gene Symbol:
CACNA1C
Gene Name:
calcium voltage-gated channel subunit alpha1 C
Gene Aliases:
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location:
Chr.12: 1969677 - 2697949 on Build GRCh38
UniGene ID:
Hs.118262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
104330
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000719.6
NP_000710.5
44
5907
NM_001129827.1
NP_001123299.1
46
6051
NM_001129829.1
NP_001123301.1
44
6030
NM_001129830.2
NP_001123302.2
45
5977
NM_001129831.1
NP_001123303.1
45
5991
NM_001129832.1
NP_001123304.1
45
5967
NM_001129833.1
NP_001123305.1
44
5964
NM_001129834.1
NP_001123306.1
44
5964
NM_001129835.1
NP_001123307.1
44
5964
NM_001129836.1
NP_001123308.1
44
5958
NM_001129837.1
NP_001123309.1
43
5931
NM_001129838.1
NP_001123310.1
43
5931
NM_001129839.1
NP_001123311.1
43
5925
NM_001129840.1
NP_001123312.1
44
5907
NM_001129841.1
NP_001123313.1
44
5907
NM_001129842.1
NP_001123314.1
44
5907
NM_001129843.1
NP_001123315.1
44
5907
NM_001129844.1
NP_001123316.1
44
5898
NM_001129846.1
NP_001123318.1
43
5874
NM_001167623.1
NP_001161095.1
44
5907
NM_001167624.2
NP_001161096.2
45
5977
NM_001167625.1
NP_001161097.1
44
6087
NM_199460.3
NP_955630.3
47
6121
XM_006719017.2
XP_006719080.1
44
5961
XM_011521020.2
XP_011519322.1
45
6036
XM_011521023.2
XP_011519325.1
45
6221
XM_017019926.1
XP_016875415.1
50
7802
XM_017019927.1
XP_016875416.1
49
7697
XM_017019928.1
XP_016875417.1
48
7484
XM_017019929.1
XP_016875418.1
48
7427
XM_017019930.1
XP_016875419.1
48
7427
XM_017019931.1
XP_016875420.1
47
7343
XM_017019932.1
XP_016875421.1
45
7328
XM_017019933.1
XP_016875422.1
46
7232
XM_017019934.1
XP_016875423.1
46
7223
XM_017019935.1
XP_016875424.1
46
7223
XM_017019936.1
XP_016875425.1
45
7205
XM_017019937.1
XP_016875426.1
45
7205
XM_017019938.1
XP_016875427.1
45
7205
XM_017019939.1
XP_016875428.1
45
7199
XM_017019940.1
XP_016875429.1
44
7172
XM_017019941.1
XP_016875430.1
44
7166
XM_017019942.1
XP_016875431.1
45
7148
XM_017019943.1
XP_016875432.1
45
7148
XM_017019944.1
XP_016875433.1
45
7148
XM_017019945.1
XP_016875434.1
45
7148
XM_017019946.1
XP_016875435.1
45
7148
XM_017019947.1
XP_016875436.1
45
7148
XM_017019948.1
XP_016875437.1
45
7148
XM_017019949.1
XP_016875438.1
45
7148
XM_017019950.1
XP_016875439.1
45
7142
XM_017019951.1
XP_016875440.1
45
7139
XM_017019952.1
XP_016875441.1
44
7115
XM_017019953.1
XP_016875442.1
44
5961
XM_017019954.1
XP_016875443.1
44
5961
XM_017019955.1
XP_016875444.1
46
7223
GenBank mRNA
AF465484.1
-
44
5859
AJ224873.1
-
44
5594
AK302698.1
-
14
1949
AY830711.1
-
43
5612
AY830712.1
-
44
5651
AY830713.1
-
44
5717
BC146846.1
-
44
5713
9056774
L29529.1
-
43
5706
L29534.1
-
44
5625
L29536.1
-
43
5520
Z26294.1
-
13
1649
Z34809.1
-
44
5594
Z34810.1
-
44
5594
Z34811.1
-
44
5651
Z34812.1
-
43
5618
Z34813.1
-
43
5618
Z34814.1
-
43
5561
Z34815.1
-
44
5594
Z34816.1
-
44
5594
Z34817.1
-
43
5597
Z34818.1
-
45
5654
Z34819.1
-
45
5678
Z34820.1
-
44
5645
Z34821.1
-
46
5738
Z34822.1
-
45
5699
Z74996.1
-
44
5651
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0