Search
Search

CLCA1| Gene Symbol: CLCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285.3 | NP_001276.2 | 8, 9 | 1494 | ||
| GenBank mRNA | AF039400.1 | - | 9, 10 | 1716 | ||
| AF127036.1 | - | 8, 9 | 1369 | |||
| AK024970.1 | - | 3, 4 | 578 | |||
| AK097057.1 | - | 8, 9 | 1227 | |||
| AK291999.1 | - | 8, 9 | 1410 | |||
| AK300858.1 | - | 4, 5 | 780 | |||
| AK314375.1 | - | 8, 9 | 1412 | |||
| AW361520.1 | - | 1, 2 | 213 | |||